Skip to Content
Merck
All Photos(1)

Documents

A7189

Sigma-Aldrich

L-Alanine Dehydrogenase from Bacillus subtilis

ammonium sulfate suspension, ≥20 units/mg protein (Lowry)

Synonym(s):

L-Alanine: NAD+ oxidoreductase (deaminating)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Bacillus subtilis

Quality Level

form

ammonium sulfate suspension

specific activity

≥20 units/mg protein (Lowry)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

L-Alanine Dehydrogenase has a N-terminal substrate-binding domain and a C-terminal NAD-binding domain.

Application

L-Alanine Dehydrogenase from Bacillus subtilis has been used in the carbon nanotube columns for H2-driven biocatalysis hydrogenation studies.
L-Alanine dehydrogenase converts L-alanine to pyruvate and ammonium. L-Alanine dehydrogenase from Bacillus subtilis may be used to study enzyme inactivation and protection .

Biochem/physiol Actions

L-Alanine Dehydrogenase is essential for sporulation in Bacillus subtilis.
L-Alanine dehydrogenase is a stereospecific dehydrogenase that catalyzes the reversible deamination of L-alanine to pyruvate and ammonium. It is important for the generation of pyruvate during sporulation. L-Alanine dehydrogenase from Bacillus subtilis has a predominately ordered kinetic mechanism in which NAD binds before L-alanine. Subsequently, ammonia, pyruvate and NADH are released in that specific order. Optimal pH for the amination reaction is 8.8-9.0, whereas it is 10-10.5 for the deamination reaction. The enzyme is inactivated by divalent metal ions and p-chloromercuribenzoate, mercuric ion being most effective. The inactivation may be reversed by L- or D-cysteine.

Unit Definition

One unit will convert 1.0 μmole of L-alanine to pyruvate and NH3 per min at pH 10.0 at 25 °C.

Physical form

Suspension in 2.4 M (NH4)2SO4 solution, pH 7.0

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H 2-Driven biocatalytic hydrogenation in continuous flow using enzyme-modified carbon nanotube columns
Zor C, et al.
Chemical Communications (Cambridge, England), 53(71), 9839-9841 (2017)
Alanine dehydrogenase (ald) is required for normal sporulation in Bacillus subtilis.
Siranosian K, et al.
Journal of Bacteriology, 175(21), 6789-6796 (1993)
Domain motions and functionally-key residues of l-alanine dehydrogenase revealed by an elastic network model
Li XY, et al.
International Journal of Molecular Sciences, 16(12), 29383-29397 (2015)
D Delforge et al.
The Journal of biological chemistry, 272(4), 2276-2284 (1997-01-24)
L-Alanine dehydrogenase from Bacillus subtilis was inactivated with two different lysine-directed chemical reagents, i.e. 2,4, 6-trinitrobenzenesulfonic acid and N-succinimidyl 3-(2-pyridyldithio)propionate. In both cases, the inactivation followed pseudo first-order kinetics, with a 1:1 stoichiometric ratio between the reagent and the enzyme
Dariel Márquez et al.
Frontiers in microbiology, 12, 695382-695382 (2021-08-24)
The γ-aminobutyric acid (GABA) shunt constitutes a conserved metabolic route generating nicotinamide adenine dinucleotide phosphate (NADPH) and regulating stress response in most organisms. Here we show that in the presence of GABA, Saccharomyces cerevisiae produces glutamate and alanine through the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service