Skip to Content
Merck
All Photos(1)

Documents

A6075

Sigma-Aldrich

Monoclonal Anti-Chicken Egg Albumin (Ovalbumin) antibody produced in mouse

clone OVA-14, ascites fluid

Synonym(s):

Anti-Albumin, chicken egg

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

OVA-14, monoclonal

mol wt

antigen 45 kDa

contains

15 mM sodium azide

species reactivity

chicken, turkey

should not react with

mouse, bovine, horse, canine, feline, pig, pigeon, rabbit, guinea pig, goat, sheep, donkey, rat, human

technique(s)

dot blot: suitable
indirect ELISA: 1:10,000 using chicken egg albumin (ovalbumin)
western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

chicken ... Alb(396197)

General description

Monoclonal Anti-Chicken Egg Albumin (Ovalbumin) (mouse IgG1 isotype) is produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.

Immunogen

chicken egg albumin.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)
Monoclonal Anti-Chicken Egg Albumin (Ovalbumin) antibody produced in mouse.

Biochem/physiol Actions

Ovalbumin (OVA) is the main component of egg white proteins in hens. OVA stimulates IgE-mediated hypersensitivity responses in individuals with egg allergy. The protein has also been used to study structure-function relationships in allergens. Thus, Monoclonal Anti-Chicken Egg Albumin (Ovalbumin) antibody can be a useful tool for the analysis of egg albumin function in mediating food allergies.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Revaud et al.
Frontiers in cellular and infection microbiology, 8, 6-6 (2018-02-10)
To define the bottlenecks that restrict antigen expression after oral administration of viral-vectored vaccines, we tracked vectors derived from the human adenovirus type 5 at whole body, tissue, and cellular scales throughout the digestive tract in a murine model of
Lindsey S Treviño et al.
Cancer prevention research (Philadelphia, Pa.), 5(2), 343-349 (2011-12-03)
Ovarian cancer is the leading cause of reproductive cancer death in U.S. women. This high mortality rate is due to the lack of early detection methods and ineffectiveness of therapy for advanced disease. Until more effective screening methods and therapies
Jarrod Predina et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(5), E415-E424 (2012-12-29)
Each year, more than 700,000 people undergo cancer surgery in the United States. However, more than 40% of those patients develop recurrences and have a poor outcome. Traditionally, the medical community has assumed that recurrent tumors arise from selected tumor
J P Zhao et al.
Journal of animal physiology and animal nutrition, 100(5), 876-883 (2016-06-09)
Three trials were performed to evaluate the association of ovalbumin (OVA) abundance in the oviduct magnum with egg production and the underlying regulatory mechanism by glucocorticoids. In trial 1, twenty Hy-Line Brown layers (56-60 weeks of age) with different combinations (n = 5/combination)
Stefan Hutzler et al.
Scientific reports, 7(1), 16892-16892 (2017-12-06)
Recombinant vaccine strain-derived measles virus (MV) is clinically tested both as vaccine platform to protect against other pathogens and as oncolytic virus for tumor treatment. To investigate the potential synergism in anti-tumoral efficacy of oncolytic and vaccine properties, we chose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service