Skip to Content
Merck
All Photos(1)

Key Documents

A5598

Sigma-Aldrich

Anti-c-Myc−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-c-Myc antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

1.8-4 mg/mL

technique(s)

western blot: 1:5,000

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... MYC(4609)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Anti-c-Myc peroxidase conjugate rabbit antibody reacts with c-Myc tagged fusion proteins. It recognizes the c-Myc tag sequence expressed at either the amino- or the carboxyl-terminus.

Specificity

N-terminal or C-terminal c-Myc (Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu) tagged fusion proteins.

Immunogen

peptide corresponding amino acids 408-425 of the human c-myc proto-oncogene, conjugated to maleimide-activated KLH through a C-terminal added cysteine residue.

Application

Antibody is suitable for immunoblotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 0.01 M sodium phosphate buffer saline, pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Du-Hwa Lee et al.
The Plant cell, 29(11), 2895-2920 (2017-11-01)
Dynamic control of protein translation in response to the environment is essential for the survival of plant cells. Target of rapamycin (TOR) coordinates protein synthesis with cellular energy/nutrient availability through transcriptional modulation and phosphorylation of the translation machinery. However, mechanisms
Yang Lu et al.
Molecular biology of the cell, 19(10), 4260-4272 (2008-08-08)
Efg1 is essential for hyphal development and virulence in the human pathogenic fungus Candida albicans. How Efg1 regulates gene expression is unknown. Here, we show that Efg1 interacts with components of the nucleosome acetyltransferase of H4 (NuA4) histone acetyltransferase (HAT)
Camilla Kreßner et al.
PloS one, 8(8), e69947-e69947 (2013-08-13)
We have recently characterized SCAI (Suppressor of Cancer Cell Invasion), a transcriptional modulator regulating cancer cell motility through suppression of MAL/SRF dependent gene transcription. We show here that SCAI is expressed in a wide range of normal human tissues and
Xiang He et al.
PLoS pathogens, 13(6), e1006347-e1006347 (2017-06-08)
Estrogen-related receptor α (ERRα) is a member of the nuclear receptor superfamily controlling energy homeostasis; however, its precise role in regulating antiviral innate immunity remains to be clarified. Here, we showed that ERRα deficiency conferred resistance to viral infection both
Gang Li et al.
Nature cell biology, 13(5), 616-622 (2011-04-19)
The circadian clock controls many metabolic, developmental and physiological processes in a time-of-day-specific manner in both plants and animals. The photoreceptors involved in the perception of light and entrainment of the circadian clock have been well characterized in plants. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service