Skip to Content
Merck
All Photos(1)

Documents

A3551

Sigma-Aldrich

Alsever′s Solution

liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Blood cell stabilizer solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

glucose: 20.5 g/L (Dextro)

shipped in

ambient

storage temp.

room temp

General description

Alsever′s solution is an isotonic, balanced salt solution. It is routinely used as an anticoagulant or blood preservative, which permits the storage of whole blood in refrigerators for approximately 10 weeks.

Application

Alsever′s Solution has been used:
  • as a preservative of whole blood collected from Meleagris gallopavo, Oreochromis niloticus, and Salmo salar
  • as a diluent for severe acute respiratory syndrome–related coronavirus-2 (SARS-CoV-2) peptide Kode Technology-modified red cells (C19-kodecytes)
  • for preserving hemolymph collected from shrimp (Litopenaeus vannamei)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radhika Nagappan et al.
Transfusion, 61(4), 1171-1180 (2021-02-17)
The Coronavirus disease 2019 (COVID-19) pandemic is having a major global impact, and the resultant response in the development of new diagnostics is unprecedented. The detection of antibodies against severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has a role in
Augustino Alfred Chengula et al.
Viruses, 11(12) (2019-12-18)
Tilapia lake virus (TiLV) is a negative-sense single-stranded RNA (-ssRNA) icosahedral virus classified to be the only member in the family Amnoonviridae. Although TiLV segment-1 shares homology with the influenza C virus PB1 and has four conserved motifs similar to
Growth performance, digestive enzymes, antioxidant activity and immune responses of Litopenaeus vannamei fed with Jania adhaerens JV Supplemented diet against Photobacterium damselae infection
Akbary P, et al.
Animal feed science and technology, 270, 114696-114696 (2020)
Yue Li et al.
Biology, 9(1) (2020-01-16)
Hypericin (Hyp) had been explored as a tumor-seeking agent for years; however, more recent studies showed its necrosis-avidity rather than cancer-seeking property. To further look into this discrepancy, we conducted an in vitro study on Hyp retention in vital and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service