Skip to Content
Merck
All Photos(1)

Key Documents

A269

Sigma-Aldrich

Anti-A2A Adenosine Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45 kDa

species reactivity

canine, human

should not react with

rat

technique(s)

immunohistochemistry (frozen sections): 1:100
immunoprecipitation (IP): suitable
western blot: 1:2000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADORA2A(135)

General description

ADORA2A is an adenosine receptor and part of the G-protein coupled receptor family of proteins. ADORA2A is involved in regulating myocardial blood flow and dopamine release in the brain.
Anti-A2A Adenosine Receptor is specific for A2A adenosine receptor adenosine receptor subtype. By immunoblotting, it reacts strongly with canine A2A. However, it does not react with rat A2A. The antibody detects A2A adenosine receptor in human hippocampus by immunohistology and may be used for immunoprecipitation.

Immunogen

Synthetic peptide (Ser-His-Gly-Asp-Met-Gly-Leu-Pro-Asp-Val-Glu-Leu-Leu-Ser-His-Glu-Leu-Lys) derived from amino acids 373-391 of the canine A2A adenosine receptor C-terminal domain.

Application

Rabbit anti-A2A adenosine receptor antibody can be used for western blot applications. The antibody can also be used for immunohistochemistry (1:100, frozen sections) and immunoprecipitation applications.

Physical form

Solution in phosphate buffered saline, containing 1 mg/ml BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liping Gao et al.
The Journal of pharmacology and experimental therapeutics, 305(2), 581-586 (2003-02-28)
Hypertension in spontaneously hypertensive rats (SHRs) is due in part to enhanced effects of vasoactive peptides on the renal vasculature. We hypothesize that the G(i) signal transduction pathway enhances renovascular responses to vasoactive peptides in SHRs more so than in
Krista M Beach et al.
Experimental eye research, 174, 40-50 (2018-05-25)
Adenosine receptor (ADOR) antagonists, such as 7-methylxanthine (7-MX), have been shown to slow myopia progression in humans and animal models. Adenosine receptors are found throughout the body, and regulate the release of neurotransmitters such as dopamine and glutamate. However, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service