A2233
AKT2, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
PKBBETA, PRKBB, RAC-BETA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
38-52 nmol/min·mg
mol wt
~85 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... AKT2(208)
Biochem/physiol Actions
AKT2 or Protein Kinase B b (PKBb) is a serine/threonine kinase that is a member of the AKT family. AKT2 like the other AKT members is activated in cells in response to diverse stimuli such as hormones, growth factors and extracellular matrix components and is involved in glucose metabolism, transcription, survival, cell proliferation, angiogenesis, and cell motility. The PI3K generates phosphatidylinositol-3, 4, 5-trisphosphate (PIP3), a lipid second messenger essential for the translocation of AKT2 to the plasma membrane where it is phosphorylated and activated by phosphoinositide-dependent kinase-1 (PDK-1).
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
P J Coffer et al.
The Biochemical journal, 335 ( Pt 1), 1-13 (1998-09-22)
While a plethora of extracellular molecules exist that modulate cellular functions via binding to membrane receptors inside the cell, their actions are mediated by relatively few signalling mechanisms. One of these is activation of phosphatidylinositol 3-kinase (PI-3K), which results in
K E Anderson et al.
Current biology : CB, 8(12), 684-691 (1998-06-25)
Protein kinase B (PKB) is involved in the regulation of apoptosis, protein synthesis and glycogen metabolism in mammalian cells. Phosphoinositide-dependent protein kinase (PDK-1) activates PKB in a manner dependent on phosphatidylinositol 3,4,5-trisphosphate (PtdIns(3,4,5)P3), which is also needed for the translocation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service