Skip to Content
Merck
All Photos(1)

Key Documents

A211800

Sigma-Aldrich

DMT-2′O-MOE-rA(bz) Phosphoramidite

Synonym(s):

5′-Dimethoxytrityl-N6-Benzoyl-3′-[(2-cyanoethyl)- (N,N-diisopropyl)]-phosphoramidite- 2′-O-(2-methoxyethyl)adenosine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C50H58N7O9P
CAS Number:
Molecular Weight:
932.01
UNSPSC Code:
41116105

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

impurities

≤0.1% single unspecified Impurity (reversed phase HPLC)
≤0.3% P(III) 100-169ppm (31P-NMR)
≤0.3 wt. % water content (Karl Fischer)
≤0.5% CET-H-Phosphonate (reversed phase HPLC)
≤0.5% DIPA-H-Phosphonate (reversed phase HPLC)
≤0.5% DMT-rA(bz)moe (reversed phase HPLC)
≤0.5% DMT-rA(bz)moe-Bz (reversed phase HPLC)
≤0.5% DMT-rA(bz)moe-amidate (reversed phase HPLC)
≤0.5% H-Phosphonat (reversed phase HPLC)
≤0.5% O3, O5-Bis-DMT-rA(bz)moe (reversed phase HPLC)
≤3 wt. % residual solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service