Skip to Content
Merck
All Photos(2)

Key Documents

A1394

Sigma-Aldrich

4-Aminophenyl α-D-mannopyranoside

≥98% (TLC)

Synonym(s):

p-aminophenyl alpha-D-mannoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO6
CAS Number:
Molecular Weight:
271.27
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (TLC)

form

powder

optical activity

[α]/D 123.00 to 135.00°, c = 9.00-11.00 mg/mL in methanol

technique(s)

thin layer chromatography (TLC): suitable

color

white to yellow cast

solubility

H2O: soluble

storage temp.

2-8°C

SMILES string

Nc1ccc(OC2OC(CO)C(O)C(O)C2O)cc1

InChI

1S/C12H17NO6/c13-6-1-3-7(4-2-6)18-12-11(17)10(16)9(15)8(5-14)19-12/h1-4,8-12,14-17H,5,13H2

InChI key

MIAKOEWBCMPCQR-UHFFFAOYSA-N

Application

4-Aminophenyl α-D-mannopyranoside is used to modify the surface of liposomes to increase uptake kinetics.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Ying et al.
Journal of controlled release : official journal of the Controlled Release Society, 141(2), 183-192 (2009-10-06)
Chemotherapy for brain glioma has been of limited value due to the inability of transport of drug across the blood-brain barrier (BBB) and poor penetration of drug into the tumor. For overcoming these hurdles, the dual-targeting daunorubicin liposomes were developed
Xue Ying et al.
Pharmacology, 87(1-2), 105-114 (2011-02-02)
To circumvent the problem of transporting anticancer drugs across the blood-brain barrier (BBB) to target brain tumors, we have previously developed dual-targeting daunorubicin liposomes modified with 4-aminophenyl-α-D-manno-pyranoside and transferrin molecules. The objective of the present study was to evaluate the
M Triggiani et al.
Journal of immunology (Baltimore, Md. : 1950), 164(9), 4908-4915 (2000-04-26)
Secretory phospholipases A2 (sPLA2s) are a group of extracellular enzymes that release fatty acids at the sn-2 position of phospholipids. Group IIA sPLA2 has been detected in inflammatory fluids, and its plasma level is increased in inflammatory diseases. To investigate
P K Das et al.
Experientia, 38(5), 629-630 (1982-05-15)
Antiserum was raised against an aminophenyl derivative of D-mannose grafted on to a liposomal surface. As characterized by immunodiffusion, quantitative precipitation and hapten inhibition, the antiserum was found to contain mannose specific antibodies in addition to antibodies against the aromatic
Sumio Chono et al.
Drug development and industrial pharmacy, 36(1), 102-107 (2009-08-07)
The effect of surface-mannose modification on aerosolized liposomal delivery to alveolar macrophages (AMs) was evaluated in vitro and in vivo. 4-Aminophenyl-α-D-mannopyranoside (Man) was used for surface-mannose modification, and mannosylated liposomes with various mannosylation rates (particle size: 1000 nm) were prepared.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service