Skip to Content
Merck
All Photos(4)

Key Documents

A1388

Sigma-Aldrich

Adenosine 5′-[γ-thio]triphosphate tetralithium salt

≥75% (HPLC), powder

Synonym(s):

ATP-γ-S, Adenosine 5′-(3-thiotriphosphate) tetralithium salt, Ado-5′-PPP[S]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12Li4N5O12P3S
CAS Number:
Molecular Weight:
546.98
Beilstein:
6265083
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥75% (HPLC)

form

powder

color

off-white

solubility

H2O: 25 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].[Li+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=S)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O12P3S.4Li/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(25-10)1-24-28(18,19)26-29(20,21)27-30(22,23)31;;;;/h2-4,6-7,10,16-17H,1H2,(H,18,19)(H,20,21)(H2,11,12,13)(H2,22,23,31);;;;/q;4*+1/p-4/t4-,6-,7-,10-;;;;/m1..../s1

InChI key

DWQFDOIBOYDYKH-KWIZKVQNSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-[γ-thio]triphosphate tetralithium salt has been used to treat the pollen grains to determine the inhibitory effect of adenosine triphosphate (ATP) or its hydrolysis product. It has also been used to select thiophosphorylate target protein.

Biochem/physiol Actions

Adenosine 5′-[γ-thio]triphosphate (ATP-γ-S) is widely used as a non-hydrolyzed ATP analogue for studies on the effects of ATP binding to ATP sites in a wide range of processes including extracellular functions. ATP- γ-S is a P2 purinergic agonist which increases the activity of Ca2+-activated K+ channels. ATP- γ-S can substitute for ATP in kinase reactions yielding thiophosphorylated proteins that are resistant to protein phosphatases and modulate the ATP-dependent steps in the binding of ubiquitin conjugates to the 26S proteasome. ATP-γ -S differentially modulates Toll-like receptor 4-mediated cell survival and death.

Other Notes

Non-hydrolyzable ATP analog

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas Peth et al.
Molecular cell, 40(4), 671-681 (2010-11-26)
Eukaryotic cells target proteins for degradation by the 26S proteasome by attaching a ubiquitin chain. Using a rapid assay, we analyzed the initial binding of ubiquitinated proteins to purified 26S particles as an isolated process at 4°C. Subunits Rpn10 and
David M Smith et al.
Cell, 144(4), 526-538 (2011-02-22)
In the eukaryotic 26S proteasome, the 20S particle is regulated by six AAA ATPase subunits and, in archaea, by a homologous ring complex, PAN. To clarify the role of ATP in proteolysis, we studied how nucleotides bind to PAN. Although
S Chrysogelos et al.
The Journal of biological chemistry, 258(20), 12624-12631 (1983-10-25)
recA protein binds to duplex DNA in the presence of Mg2+ and adenosine 5'-O-(3-thiotriphosphate) forming a stiff nucleoprotein filament with a distinct axial repeat which contains 17 +/- 1 base pairs and spans 8-9 nm along the fiber (Di Capua
Xiang Chen et al.
STAR protocols, 2(1), 100278-100278 (2021-02-02)
The 26S proteasome is specialized for regulated protein degradation. It is formed by a regulatory particle (RP) that recognizes ubiquitinated substrates and caps a hollow cylindrical core particle (CP) where substrates are proteolyzed. Structural heterogeneity caused by dynamics makes it
Kana Harada et al.
Journal of neurochemistry, 116(6), 1138-1147 (2011-01-08)
The survival and death rates of inflammatory cells directly control their number and are substantially associated with the degree of inflammation. Microglia, key players in neuroinflammation, often cause excessive reactions implicated in neurological diseases. However, the mechanisms that determine microglial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service