Skip to Content
Merck
All Photos(1)

Key Documents

A0802

Sigma-Aldrich

Adipic acid dihydrazide–Agarose

saline suspension

Synonym(s):

Adipic acid dihydrazide agarose beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

biological source

plant

form

saline suspension

extent of labeling

≥8 μmol per mL

technique(s)

microbe id | metabolite detection: suitable

matrix activation

cyanogen bromide

matrix spacer

11 atoms (when ligands (aldehydes, carboxylic acids) are coupled through free hydrazide groups)

suitability

suitable for chromatography

storage temp.

2-8°C

Related Categories

Application

Adipic acid dihydrazide Agarose can be used in proteomics and protein chromatography. It has been utilized in research for purifying and identifying the fungal phytotoxin Fusicoccin (FC), identifying RNA binding proteins that interact with RNA cis-elements, and enzyme purification such as peroxisomes from guinea pig liver.

Physical form

Suspension in 0.5 M NaCl containing preservative.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruben Hovhannisyan et al.
BioTechniques, 46(2), 95-98 (2009-03-26)
Use of RNA affinity chromatography is commonly used to identify RNA binding proteins that interact with specific RNA cis-elements that function in post-transcriptional gene regulation. These purifications can be complicated by residual RNase activity in cellular extracts that can degrade
S C Datta et al.
The Journal of biological chemistry, 265(14), 8268-8274 (1990-05-15)
The peroxisomal acyl/alkyl dihydroxyacetone-phosphate reductase (EC 1.1.1.101) was solubilized and purified 5500-fold from guinea pig liver. The enzyme could be solubilized by detergents only at high ionic strengths in presence of the cosubstrate NADPH. Peroxisomes, isolated from liver by a
M Caputi et al.
The EMBO journal, 18(14), 4060-4067 (1999-07-16)
Splicing of the human immunodeficiency virus type 1 (HIV-1) pre-mRNA must be inefficient to provide a pool of unspliced messages which encode viral proteins and serve as genomes for new virions. Negative cis-regulatory elements (exonic splicing silencers or ESSs) are
Alice Tianbu Zhang et al.
Journal of cell science, 124(Pt 12), 2058-2069 (2011-05-26)
Non-coding Y RNAs are required for the initiation of chromosomal DNA replication in mammalian cells. It is unknown how they perform this function or if they associate with a nuclear structure during DNA replication. Here, we investigate the association of
Abrahem Abrahem et al.
Nucleic acids research, 36(16), 5201-5211 (2008-08-07)
Although RNA polymerases (RNAPs) are able to use RNA as template, it is unknown how they recognize RNA promoters. In this study, we used an RNA fragment derived from the hepatitis delta virus (HDV) genome as a model to investigate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service