Skip to Content
Merck
All Photos(2)

Key Documents

A0521

Sigma-Aldrich

α-Amylase from human saliva

greener alternative

Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein

Synonym(s):

1,4-α-D-Glucan-glucanohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

type

Type IX-A

Quality Level

form

lyophilized powder

specific activity

1,000-3,000 units/mg protein

composition

Protein, 5.0-15.0% biuret

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

UniProt accession no.

greener alternative category

storage temp.

−20°C

Gene Information

human ... AMY1A(276)

Looking for similar products? Visit Product Comparison Guide

General description

α-amylase is the main secretory protein constituent of saliva. It contains 496 amino acids and has two isoforms, glycosylated and non-glycosylated. α-amylase gene is located on human chromosome 1p21.1. α-amylase is mainly produced in pancreas and saliva.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in starch ethanol research. For more information see the article in biofiles.

Application

α-amylase from human saliva has been used to test α-amylase inhibitors using HPLC (high performance liquid chromatography). It also has been used to study the digestibility of human milk oligosaccharides (HMO).

Biochem/physiol Actions

α-amylase catalyses the hydrolysis of α-1,4 glycosidic linkage in oligosaccharides. It plays a crucial role in the initial digestion of starch, glycogen and polysaccharides. Elevated levels of amylase in serum is linked to acute pancreatitis.

Unit Definition

One unit will liberate 1.0 mg of maltose from starch in 3 min at pH 6.9 at 20 °C.

Physical form

Lyophilized powder containing (NH4)2SO4 and sodium citrate

Preparation Note

Chromatographically purified

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Copy number polymorphism of the salivary amylase gene: implications in human nutrition research
Santos JL, et al.
Lifestyle Genomics, 5(3), 117-131 (2012)
Structure of human salivary alpha-amylase crystallized in a C-centered monoclinic space group
Fisher SZ, et al.
Acta Crystallographica Section F, Structural Biology and Crystallization Communications, 62(2), 88-93 (2006)
M J Gnoth et al.
The Journal of nutrition, 130(12), 3014-3020 (2000-12-09)
In examining the functional aspects of human milk oligosaccharides (HMO), it is not known whether they are digested during the passage through the infant's gastrointestinal tract. HMO were prepared from individual milk samples (n = 6) and separated into neutral
Maria Alberta Araújo et al.
Journal of biomedical materials research. Part A, 94(3), 720-729 (2010-03-13)
This study aims at evaluating the influence of enzymatic degradation solution on the surface morphology and thermal properties of a poly(ethylene-vinyl alcohol) copolymer-corn starch thermoplastic blend (SEVA-C), as a function of immersion time. To perform this study, three different batches
HPLC method for measurement of human salivary alpha-amylase inhibition by aqueous plant extracts
Takacs I, et al.
PLoS ONE, 68(2), 127-136 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service