Skip to Content
Merck
All Photos(1)

Key Documents

960D

Sigma-Aldrich

Permanent Red Chromogen Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

description

For In Vitro Diagnostic Use in Select Regions

Quality Level

form

aqueous solution

manufacturer/tradename

Cell Marque

packaging

kit of (containing 0.7 mL each of 3 reagents + 30 mL buffer (960D-10))
kit of (containing 2.3 mL each of 3 reagents + 100 mL buffer (960D-20))

technique(s)

immunohistochemistry: suitable

storage temp.

2-8°C

General description

The permanent Red Chromogen kit is intended for laboratory use to indentify by light microscopy target antigens in formalin fixed paraffin embedded tissue or frozen tissue when used in conjunction with antibodies and detection reagents using Alkaline Phosphatase enzyme in the IHC staining process.Cell Marque offers and recommends the use of its Alkaline Phosphatase labeled detection systems.When in the presence of Alkaline Phosphatase, Permanent Red Chromogen produces a non-fading, bright red precipitate that can be readily visualized using light microscopy. The precipitate is insoluble in organic solvents and can be coverslipped with permanent mounting media.

Quality

United States - IVD
Canada - RUO
European Union - IVD
Japan - RUO

Physical form

Ready to use reagent

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service