Skip to Content
Merck
All Photos(1)

Key Documents

88276

Sigma-Aldrich

Poly(ethylene glycol)

BioUltra, 3,350

Synonym(s):

Polyethylene glycol, Polyethylene oxide, Polyoxyethylene, PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Quality Level

product line

BioUltra

form

powder

mol wt

Mr 3000-3700

impurities

insoluble matters, passes filter test

pH

5.5-7.0 (25 °C, 50 mg/mL in H2O)

solubility

H2O: 50 mg/mL, clear, colorless

anion traces

chloride (Cl-): <50 mg/kg
sulfate (SO42-): <50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

50 mg/mL in H2O

UV absorption

λ: 260 nm Amax: 0.06
λ: 280 nm Amax: 0.03

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is a linear polyether of ethylene glycol (ethane-1,2-diol).[1] It is soluble in water, benzene, ethanol, acetonitrile, and dichloromethane. However, it is insoluble in diethyl ether and hexane.[2] This non-ionic polymer[3] can exist in the monomer and aqueous phases.[4]

Application

Poly(ethylene glycol) has been used:

  • to grow the crystals of HLA-A∗02:01-N222-230 LLLDRLNQL (pHLA) complex[5]
  • in the fabrication of ultrathin, free-standing, and porous polymer membranes for retinal tissue engineering[6]
  • to grow the crystals of the C2c2-crRNA complex[7]
  • to grow the crystals of the apo-C2c2 thermolysin-resistant core[7]

Biochem/physiol Actions

Poly(ethylene glycol) (PEG) is used in several pharmaceutical applications.[2] This polymer occurs as a preservative and is used in biomedical applications like drug delivery systems, biosensing, and tissue engineering. PEG is also used in cosmetic ointments and creams. It is used as a binding and dispersing agent in chemical and other industries.[8]

Features and Benefits

  • Environment friendly[4]
  • Biocompatible[4]
  • Unique solvent solution[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang Liu et al.
Cell, 168(1-2), 121-134 (2017-01-14)
C2c2, the effector of type VI CRISPR-Cas systems, has two RNase activities-one for cutting its RNA target and the other for processing the CRISPR RNA (crRNA). Here, we report the structures of Leptotrichia shahii C2c2 in its crRNA-free and crRNA-bound
Polymers for 3D bioprinting
Beata K
Journal of Separation Science, 337-349 (2022)
Polymers and polymeric hybrids for targeted drug delivery
Islam R and Fang J
Journal of Separation Science, 303-322 (2022)
Application of compatibilized polymer blends in biomedical fields
Zarrintaj P, et al.
Journal of Separation Science, 511-537 (2020)
Novel method for the fabrication of ultrathin, free-standing and porous polymer membranes for retinal tissue engineering
Tan EYS, et al.
Journal of materials chemistry. B, 5(28), 5616-5622 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service