Skip to Content
Merck
All Photos(1)

Documents

86524

Sigma-Aldrich

CM-Dextran sodium salt

BioXtra

Synonym(s):

CM-D, Carboxymethyl-dextran sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

product line

BioXtra

Quality Level

form

powder

impurities

≤10% water

color

white

solubility

H2O: 10 mg/mL, clear, colorless

anion traces

chloride (Cl-): ≤5500 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤500 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤500 mg/kg
Mg: ≤100 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤10 mg/kg
Zn: ≤10 mg/kg

λ

10 mg/mL in H2O

UV absorption

λ: 260 nm Amax: ≤0.03
λ: 280 nm Amax: ≤0.04

Application

Carboxymethyl-dextran (CMD) may be used as a matrix component to prepare surfaces for cell adherence and culture and in the construction of polymer membranes used in the development of biomaterials/biostructures for applications such as drug delivery.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

in vitromorphogenesis of PANC-1 cells into islet-like aggregates using RGD-covered dextran derivative surfaces.
Dubiel EA, Kuehn C, Wang R, Vermette P.
Colloids and Surfaces, B: Biointerfaces, 89, 117-125 (2012)
Carina Kuehn et al.
Acta biomaterialia, 8(2), 619-626 (2011-11-17)
Rat insulinoma cells (INS-1), an immortalized pancreatic beta cell line, were cultured on low-fouling carboxymethyl-dextran (CMD) layers bearing fibronectin, the tripeptide Arg-Gly-Asp (RGD) or CDPGYIGSR, a laminin nonapeptide. INS-1 cells were non-adherent on CMD and RGE but adhered to fibronectin-
Characterization of Chitosan-Carboxymethyl Dextran Nanoparticles as a Drug Carrier and as a Stimulator of Mouse Splenocytes.
Lin YS, Radzi R, Morimoto M, et al.
Journal of Biomaterials Science. Polymer Edition, doi: 10-doi: 10 (2011)
Wang Xiao et al.
The journal of physical chemistry. B, 115(46), 13796-13802 (2011-10-25)
In this study, photoswitchable microcapsules were fabricated based on host-guest interactions between α-cyclodextrin (α-CD) and azobenzene (Azo). Carboxymethyl dextran-graft-α-CD (CMD-g-α-CD) and poly(acrylic acid) N-aminododecane p-azobenzeneaminosuccinic acid (PAA-C(12)-Azo) were assembled layer by layer on CaCO(3) particles. α-CD-rhodamine B (α-CD-RhB), used as
Afanasy V Lunin et al.
Journal of colloid and interface science, 541, 143-149 (2019-01-28)
Exploration of novel types of iron oxide nanoparticles as well as novel versatile ways to prepare them in a controlled manner keeping in mind necessity of narrow size distributions and high colloidal and chemical stability is an important task for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service