Skip to Content
Merck
All Photos(1)

Documents

81709

Sigma-Aldrich

L-Proline

≥99.5% (NT), suitable for cell culture, BioUltra

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80810
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Proline, BioUltra, ≥99.5% (NT)

product line

BioUltra

Assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D −85.0±1.0°, c = 5% in H2O

technique(s)

cell culture | mammalian: suitable

impurities

insoluble matter, passes filter test
≤0.3% foreign amino acids

ign. residue

≤0.1%

loss

≤0.1% loss on drying, 20 °C (HV)

color

colorless to white

pH

6.0-7.0 (25 °C, 1 M in H2O)

mp

228 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
NH4+: ≤20 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.05

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis and in silico inhibitory action studies of azo-anchored imidazo[4,5-b]indole scaffolds against the COVID-19 main protease (M(pro)).: This publication discusses the synthesis of novel compounds using L-Proline and their potential as inhibitors against COVID-19, contributing to the ongoing search for effective treatments (Geedkar et al., 2024).

Other Notes

Protects enzymes against heat inactivation; Growth requirement of various microorganisms

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CRC Handbook of Microbiology, 4, 1-1 (1974)
L.G. Paleg et al.
Australian Journal of Plant Physiology, 8, 107-107 (1981)
Marie Couturier et al.
The Journal of biological chemistry, 288(20), 14624-14635 (2013-04-06)
The microbial deconstruction of the plant cell wall is a key biological process that is of increasing importance with the development of a sustainable biofuel industry. The glycoside hydrolase families GH5 (PaMan5A) and GH26 (PaMan26A) endo-β-1,4-mannanases from the coprophilic ascomycete
Michael P Manns et al.
Gastroenterology, 147(2), 366-376 (2014-04-15)
MK-5172 is an inhibitor of the hepatitis C virus (HCV) nonstructural protein 3/4A protease; MK-5172 is taken once daily and has a higher potency and barrier to resistance than licensed protease inhibitors. We investigated the efficacy and tolerability of MK-5172
David Saadoun et al.
Annals of the rheumatic diseases, 73(5), 831-837 (2013-04-23)
The standard-of-care treatment of patients with hepatitis C virus (HCV)-mixed cryoglobulinemia (MC) vasculitis includes pegylated interferon α (PegIFN)-α plus ribavirin and/or rituximab. About 30-40% of patients are non-responders or relapsers to such combination. To analyse the safety and efficacy of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service