Skip to Content
Merck
All Photos(1)

Key Documents

77087

Sigma-Aldrich

ω-Transaminase, Aspergillus fumigatus

recombinant, expressed in E. coli, ≥0.20 U/mg

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

powder

specific activity

≥0.20 U/mg

storage temp.

−20°C

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Unit Definition

1 U corresponds to the amount of enzyme which releases 1 μmol acetophenone per minute at 30°C. (R(−)-α-methyl-benzylamine as substrate).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enzymatic studies on the metabolism of beta-alanine.
O HAYAISHI et al.
The Journal of biological chemistry, 236, 781-790 (1961-03-01)
Lack of stringent stereospecificity in the inactivation of pyridoxal phosphate-dependent enzymes by suicide-substrates.
C Danzin et al.
Progress in clinical and biological research, 144A, 377-385 (1984-01-01)
Hyungdon Yun et al.
Applied and environmental microbiology, 70(4), 2529-2534 (2004-04-07)
Alcaligenes denitrificans Y2k-2 was obtained by selective enrichment followed by screening from soil samples, which showed omega-amino acid:pyruvate transaminase activity, to kinetically resolve aliphatic beta-amino acid, and the corresponding structural gene (aptA) was cloned. The gene was functionally expressed in
J S Shin et al.
Bioscience, biotechnology, and biochemistry, 65(8), 1782-1788 (2001-10-02)
Microorganisms that are capable of (S)-enantioselective transamination of chiral amines were isolated from soil samples by selective enrichment using (S)-alpha-methyl-benzylamine ((S)-alpha-MBA) as a sole nitrogen source. Among them, Klebsiella pneumoniae JS2F, Bacillus thuringiensis JS64, and Vibrio fluvialis JS17 showed good
K Yonaha et al.
The Journal of biological chemistry, 267(18), 12506-12510 (1992-06-25)
The complete amino acid sequence of bacterial omega-amino acid:pyruvate aminotransferase (omega-APT) was determined from its primary structure. The enzyme protein was fragmented by CNBr cleavage, trypsin, and Staphylococcus aureus V8 digestions. The peptides were purified and sequenced by Edman degradation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service