Skip to Content
Merck
All Photos(1)

Key Documents

76183

Sigma-Aldrich

6-O-Palmitoyl-L-ascorbic acid

BioXtra, ≥99.0% (RT)

Synonym(s):

Ascorbyl palmitate, L-Ascorbic acid 6-hexadecanoate, L-Ascorbyl palmitate, Ascorbic acid 6-palmitate, NSC 402451

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H38O7
CAS Number:
Molecular Weight:
414.53
Beilstein:
96552
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic

product line

BioXtra

Assay

≥99.0% (RT)

form

powder

optical activity

[α]20/D +23±1°, c = 1% in ethanol

ign. residue

≤0.2% (as SO4)

loss

≤0.2% loss on drying

color

white to faint yellow

mp

113-116 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

InChI

1S/C22H38O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(24)28-16-17(23)21-19(25)20(26)22(27)29-21/h17,21,23,25-26H,2-16H2,1H3/t17-,21+/m0/s1

InChI key

QAQJMLQRFWZOBN-LAUBAEHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

6-O-Palmitoyl-L-ascorbic acid is a lipid soluble ester of ascorbic acid. Ascorbic acid 6-palmitate is a lipophilic derivative of ascorbic acid.

Application

6-O-Palmitoyl-L-ascorbic acid has been used to study the effects of dietary supplement in Solea senegalensis larvae.

Biochem/physiol Actions

6-O-Palmitoyl-L-ascorbic acid possesses metastasis inhibitory and anti-tumor activity. Ascorbyl palmitate exhibits antioxidant activity by protecting the cell membranes from oxidative damage.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensation of L-ascorbic acid and medium-chain fatty acids by immobilized lipase in acetonitrile with low water content
Watanabe Y, et al.
Food Science and Technology Research, 5(2), 188-192 (1999)
Oxidation and oligomerization of ethyl linoleate under the influence of the combination of ascorbic acid 6-palmitate/iron-2-ethylhexanoate
Micciche F, et al.
Applied Catalysis A: General, 297(2), 174-181 (2006)
Effect of dietary vitamin C level during early larval stages in Senegalese sole (Solea senegalensis)
Jimenez-Fernandez E, et al.
Aquaculture (Amsterdam, Netherlands), 443(2), 65-76 (2015)
The hydroxyl free radical reactions of ascorbyl palmitate as measured in various in vitro models
Perricone N, et al.
Biochemical and biophysical research communications, 262(3), 661-665 (1999)
L Capretto et al.
International journal of pharmaceutics, 440(2), 195-206 (2012-08-14)
The current paper reports the production of polymeric micelles (PMs), based on pluronic block-copolymers, as drug carriers, precisely controlling the cellular delivery of drugs with various physico-chemical characteristics. PMs were produced with a microfluidic platform to exploit further control on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service