Skip to Content
Merck
All Photos(1)

Key Documents

75903

Sigma-Aldrich

Guanosine

≥97.0% (HPLC)

Synonym(s):

9-(β-D-Ribofuranosyl)guanine, Guanine-9-β-D-ribofuranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O5
CAS Number:
Molecular Weight:
283.24
Beilstein:
625911
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥97.0% (HPLC)

form

powder

optical activity

[α]20/D -73±2°, c = 1.5% in 1 M NaOH

impurities

≤6.0% water

mp

250 °C (dec.) (lit.)

SMILES string

[H]O[H].NC1=Nc2c(ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H13N5O5/c11-10-13-7-4(8(19)14-10)12-2-15(7)9-6(18)5(17)3(1-16)20-9/h2-3,5-6,9,16-18H,1H2,(H3,11,13,14,19)/t3-,5-,6-,9-/m1/s1

InChI key

NYHBQMYGNKIUIF-UUOKFMHZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Guanosine is a purine nucleoside that upon sequential phosphoylation (kinases) forms GTP which is used by RNA polymerases to synthesis RNA(s). GTP is also important in a wide range of GTPase-dependent cell signaling processes. Guanosine is used in selective matricies such a guanosine gels (G-gels) for use in capillary electrophoresis chiral selection and other applications.

Other Notes

Currently available only in Europe.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoran D Matović et al.
Journal of inorganic biochemistry, 121, 134-144 (2013-02-05)
Novel square-planar palladium(II) complexes with O-N-N-O-type ligands H4mda (H4mda=malamido-N,N'-diacetic acid) and H4obp (H4obp=oxamido-N,N'-di-3-propionic acid) were prepared and characterized. The ligands coordinate to the palladium(II) ion via two pairs of deprotonated ligating atoms with square chelation. A four coordinate, square-planar geometry
Jeffery T Davis et al.
Chemical Society reviews, 36(2), 296-313 (2007-02-01)
Nature's use of a simple genetic code to enable life's complex functions is an inspiration for supramolecular chemistry. DNA nucleobases carry the key information utilizing a variety of cooperative and non-covalent interactions such as hydrophobic, van der Waals, pi-pi stacking
Cynthia J Burrows et al.
Environmental health perspectives, 110 Suppl 5, 713-717 (2002-11-12)
In vitro work in this laboratory has identified new DNA lesions resulting from further oxidation of a common biomarker of oxidative damage, 8-oxo-7,8-dihydroguanine (OG). The major product of oxidation of OG in a nucleoside, nucleotide, or single-stranded oligodeoxynucleotide using metal
Zhe Li et al.
Nucleic acids research, 42(6), 4068-4079 (2013-12-29)
G-quadruplex-forming oligonucleotides containing modified nucleotide chemistries have demonstrated promising pharmaceutical potential. In this work, we systematically investigate the effects of sugar-modified guanosines on the structure and stability of a (4+0) parallel and a (3+1) hybrid G-quadruplex using over 60 modified
Thomas Gonatopoulos-Pournatzis et al.
The Biochemical journal, 457(2), 231-242 (2013-12-21)
The 7mG (7-methylguanosine cap) formed on mRNA is fundamental to eukaryotic gene expression. Protein complexes recruited to 7mG mediate key processing events throughout the lifetime of the transcript. One of the most important mediators of 7mG functions is CBC (cap-binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service