Skip to Content
Merck
All Photos(2)

Documents

63705

Sigma-Aldrich

Sodium 2-mercaptoethanesulfonate

BioXtra, ≥98.0% (RT)

Synonym(s):

2-Mercaptoethanesulfonic acid sodium salt, Coenzyme M sodium salt, HS-CoM Na, MESNA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2SO3Na
CAS Number:
Molecular Weight:
164.18
Beilstein:
3657828
EC Number:
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NA.25

product line

BioXtra

Quality Level

Assay

≥98.0% (RT)

loss

≤2.0% loss on drying, 20 °C (HV)

cation traces

Ca: ≤150 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤100 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[O-]S(=O)(=O)CCS

InChI

1S/C2H6O3S2.Na/c3-7(4,5)2-1-6;/h6H,1-2H2,(H,3,4,5);/q;+1/p-1

InChI key

XOGTZOOQQBDUSI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

2-Mercaptoethanesulfonic acid sodium salt (Coenzyme M sodium salt, MESNA) is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.

Other Notes

Biosynthesis of coenzyme M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.H. White
Biochemistry, 24, 6487-6487 (1985)
Jun-ichi Nishigaki et al.
Inorganic chemistry, 51(6), 3690-3697 (2012-03-10)
Divalent and trivalent nickel complexes of 1,4,8,11-tetraazacyclotetradecane, denoted as cyclam hereafter, coordinated by methyl coenzyme M (MeSCoM(-)) and coenzyme M (HSCoM(-)) have been synthesized in the course our model studies of methyl coenzyme M reductase (MCR). The divalent nickel complexes
Janelle B Perkins et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(24), 7743-7753 (2011-10-27)
To determine the maximum tolerated dose (MTD) of topotecan in combination with ifosfamide, mesna, and etoposide (TIME), followed by autologous hematopoietic cell transplant (HCT), in patients with chemotherapy-refractory malignancies. Patients were treated with (in mg/m(2)/d) ifosfamide 3,333, mesna 3,333, and
Muhammad Faizan Nazar et al.
Journal of colloid and interface science, 363(2), 490-496 (2011-08-27)
A viable cost-effective approach employing mixtures of non-ionic surfactants Triton X-114/Triton X-100 (TX-114/TX-100), and subsequent cloud point extraction (CPE), has been utilized to concentrate and recycle inorganic nanoparticles (NPs) in aqueous media. Gold Au- and palladium Pd-NPs have been pre-synthesized
Twalib Ngoma et al.
British journal of haematology, 158(6), 749-762 (2012-08-01)
Prior to the introduction of the International Network for Cancer Treatment and Research (INCTR) protocol INCTR 03-06, survival of patients with Burkitt lymphoma at four tertiary care centres in equatorial Africa was probably no more than 10-20%. The results reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service