Skip to Content
Merck
All Photos(1)

Key Documents

61354

Sigma-Aldrich

Kollicoat® MAE 100 P

43.2-47.6% methacrylic acid basis (calc. on dried substance)

Synonym(s):

Methacrylic acid-ethyl acrylate copolymer (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.25

Quality Level

Assay

43.2-47.6% methacrylic acid basis (calc. on dried substance)

impurities

≤100 ppm residual monomer (methacrylic acid and ethyl acrylate) (HPLC)
≤20 ppm heavy metals (verified on random samples only)

ign. residue

0.5-3.0% (verified on random samples only)

loss

≤5.0% loss on drying

viscosity

≤100 mPa.s(23 °C) (RV2, 100 PRM, type B)

cation traces

As: ≤2 ppm (verified on random samples only)

SMILES string

O(CC)C(=O)C=C.OC(=O)C(=C)C

InChI

1S/C5H8O2.C4H6O2/c1-3-5(6)7-4-2;1-3(2)4(5)6/h3H,1,4H2,2H3;1H2,2H3,(H,5,6)

InChI key

GDCRSXZBSIRSFR-UHFFFAOYSA-N

Application

Kollicoat MAE can be employed as film coatings (intelligent surfaces). Kollicoat polymers can be used with all standard coating equipment and are cost-effective, delivering maximum quality in terms of function, stability, and appearance. This research grade product is intended for use in R&D and development only.
Kollicoat® MAE 100 P has been used for the production of encapsulated recombinant cytokine (recombinant tumor necrosis factor α).

Analysis Note

solubility: appearance and solubility of a film: clear, soluble in phosphate buffer solution

Legal Information

Kollicoat is a registered trademark of BASF SE

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recombinant TNFa as oral vaccine adjuvant protects European sea bass against vibriosis: insights into the role of the CCL25/CCR9 axis.
Galindo-Villegas J
Fish & Shellfish Immunology, 35, 1260-1260 (2013)
The hygroscopicity of moisture barrier film coatings.
Mwesigwa E, Buckton G, Basit AW.
Drug Devel. Ind. Pharm., 31, 959-968 (2005)
H Kranz et al.
International journal of pharmaceutics, 380(1-2), 112-119 (2009-07-28)
The major aim of this study was to identify an efficient tool to adjust drug release patterns from aqueous and organic ethylcellulose (a gastrointestinal insoluble polymer) coated pellets and to evaluate the long term stability of the film coatings. Drug
J Bajdik et al.
Die Pharmazie, 61(10), 887-888 (2006-10-31)
In the present study the effects of the additives and of storage on the deformation of films prepared from methacrylic acid/ethyl acrylate copolymer with and without polysorbate 80 were evaluated. The films containing polysorbate 80 revealed a longer deformation process
Strain Screening, Fermentation, Separation, and Encapsulation for Production of Nattokinase Functional Food.
Wei X, Luo M, Xie Y, et al.
Applied Biochemistry and Biotechnology (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service