Skip to Content
Merck
All Photos(2)

Key Documents

602-05A

Sigma-Aldrich

Hair Follicle Dermal Papilla Cells: HFDPC, adult

Synonym(s):

HFDPC, HFDPC cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human scalp (hair follicle papilla)

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

morphology

mesenchymal

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

Alzheimer′s disease

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Human Hair Follicle Dermal Papilla Cells (HFDPC) are mesenchymal cells isolated from the hair papilla of normal human scalp hair follicles. Hair papilla, in the adult hair follicle, plays a crucial role in the dermal-epidermal interactions that control hair production and events of the hair growth cycle. HFDPC can therefore be used for studying cellular mechanisms organizing the hair follicle, and for the development and evaluation of hair growth products.

HFDPC have been used to demonstrate that IL-1α affects dermal papilla activity by decreasing androgen receptor expression and stimulating secretion of KFG, VEGF, IL-8 and GM-CSF (Boivin, 2006). They were also used to investigate how gene expression and cell proliferation are affected by testosterone analogues (Reiter, 2008), potential hair growth enhancing agents (Sasajima, 2008) and microbial metabolites (Takigawa, 2012). Using HFDPC, along with Human Epidermal Keratinocytes and Human Bronchial Epithelial Cells, scientists also studied mechanisms of cellular senescence, and develop ed methodology to extend cellular life span and immortalize the cells by inhibiting p16INK4a and introducing hTERT (Haga, 2007). Finally, HFDPC have been used to discover the link between the androgen-induced alopecia and circadian rhythms, mediated by a “clock gene” transcription factor BMAL1 (Watabe, 2013).

Cell Line Origin

Scalp

Application

dermatalogical interactions, hair production, induction and maintenance of hair growth, hair follicle organization, development and evaluation of hair growth products, effects and secretion of growth factors and cytokines, gene expression, cell proliferation, cell senescence, cell lifespan extension, in vitro screening of androgen blocking agents, reservoir of multi-potent stem cells, skin reconstruction, wound healing, signal transduction pathways, skin composites and substitutes, hair follicle neogenesis

Components

Cell Basal Medium containing 20% FBS & 5% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Cell Basal Medium containing 20% FBS & 5% DMSO
  • Can be cultured at least 6 doublings

Subculture Routine

Please refer to the HFDPC Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service