Skip to Content
Merck
All Photos(1)

Key Documents

59815

Sigma-Aldrich

1-(4-Isothiocyanatobenzyl)ethylenediamine-N,N,N′,N′-tetraacetic acid

~90% (HPLC)

Synonym(s):

Isothiocyanatobenzyl-EDTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N3O8S
CAS Number:
Molecular Weight:
439.44
Beilstein:
8363400
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Assay

~90% (HPLC)

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)CN(CC(Cc1ccc(cc1)N=C=S)N(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C18H21N3O8S/c22-15(23)7-20(8-16(24)25)6-14(21(9-17(26)27)10-18(28)29)5-12-1-3-13(4-2-12)19-11-30/h1-4,14H,5-10H2,(H,22,23)(H,24,25)(H,26,27)(H,28,29)

InChI key

VHMWJVAFPCGUTJ-UHFFFAOYSA-N

Application

Isothiocyanatobenzyl-EDTA is a bifunctional chelator used in bioconjugation chemistry to label primary amine containing molecules and proteins preferentially at their N-terminals.

Other Notes

Amine-reactive probe, useful for antibody chelate labeling

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Shah et al.
Bioconjugate chemistry, 7(3), 283-289 (1996-05-01)
The synthesis of a C-5 modified uridine phosphoramidite which contains a primary amino group protected with Fmoc is described. During cleavage and deprotection of chemically synthesized RNA, the Fmoc protecting group is removed to yield a free amino group at
S V Deshpande et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 31(2), 218-224 (1990-02-01)
The metabolism of radiolabeled antibodies is important for radioimmunoimaging and therapy. The loss of indium-111 (111In) from the chelate can pose problems in imaging and increases the radiation dose to normal tissues. We have evaluated the loss in vivo of
Z Yao et al.
Nuclear medicine and biology, 22(2), 199-203 (1995-02-01)
Tn antigen is a glycosylated tumor associated antigen and a murine monoclonal antibody, MLS128, has been identified to react with it. The potential of MLS128 for the radioimmunoimaging of colorectal cancer was studied. MLS128 was labeled with radioiodine by the
T M Rana et al.
Bioconjugate chemistry, 1(5), 357-362 (1990-09-01)
Conjugates of monoclonal antibodies with drugs, toxins, radionuclides, and other agents are in widespread use in therapeutic trials and as clinical research tools. The characterization of these immunoconjugates generally does not include determining the individual sites at which such agents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service