Skip to Content
Merck
All Photos(1)

Key Documents

57858

Sigma-Aldrich

Sodium iodoacetate

BioUltra, ≥99.5% (NT)

Synonym(s):

Iodoacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2COONa
CAS Number:
Molecular Weight:
207.93
Beilstein:
4009322
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

product line

BioUltra

Quality Level

Assay

≥99.5% (NT)

impurities

insoluble matter, passes filter test
≤0.5% water

mp

208 °C (dec.) (lit.)

solubility

methanol: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤500 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

absorption

cut-off at 335 nm in methanol at 0.1 M

storage temp.

−20°C

SMILES string

[Na+].[O-]C(=O)CI

InChI

1S/C2H3IO2.Na/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1

InChI key

AGDSCTQQXMDDCV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for the modification of cysteine residues in proteins. Used in protein sequencing (including by Sanger in sequencing bovine insulin) to prevent oxidation of -SH sidechains.
Reagent for the modification of cysteine residues in proteins. Used to induce animal osteoarthritis model system, which mimics both symptoms and histopathology of the human disease.

Other Notes

Sulfhydryl reagent

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

406.4 °F

Flash Point(C)

208 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Gagaoua et al.
Journal of agricultural and food chemistry, 63(30), 6799-6809 (2015-07-15)
Many studies on color biochemistry and protein biomarkers were undertaken in post-mortem beef muscles after ≥24 hours. The present study was conducted on Longissimus thoracis muscles of 21 Blond d'Aquitaine young bulls to evaluate the relationships between protein biomarkers present
F.R. Gurd
Methods in Enzymology, 11, 532-532 (1973)
J L Leonard et al.
Biochimica et biophysica acta, 787(2), 122-130 (1984-06-14)
Pretreatment of renal iodothyronine 5'-deiodinase with sulfhydryl reagents, iodoacetate, iodoacetamide and N-alkylmaleimides, results in irreversible loss of catalytic activity. Iodoacetate and iodoacetamide were the most potent inhibitors, being 100- to 1000-times more potent than N-alkylmaleimides. Iodoacetate and iodoacetamide inactivation followed
Joon Shik Yoon et al.
American journal of physical medicine & rehabilitation, 94(8), 644-648 (2014-11-22)
The aim of this study was to investigate the potential of using phase-contrast radiography to detect early changes in articular cartilage in a mouse model of osteoarthritis. An iodoacetate-induced mouse model of osteoarthritis was used. Mice with iodoacetate-induced osteoarthritis were
K Nakyinsige et al.
Meat science, 98(4), 701-708 (2014-08-05)
This study assessed the effect of gas stunning which has not been conducted until now in comparison with slaughter without stunning on the welfare and meat quality of rabbits. Eighty male New Zealand White rabbits were divided into two groups

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service