Skip to Content
Merck
All Photos(1)

Key Documents

56901

Sigma-Aldrich

Geranyl monophosphate lithium salt

≥95.0% (TLC)

Synonym(s):

(E)-3,7-Dimethyl-2,6-octadien-1-ol dihydrogen phosphate lithium salt, (E)-3,7-Dimethyl-2,6-octadien-1-yl phosphate lithium salt, Geranyl phosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O4P · xLi+
CAS Number:
Molecular Weight:
234.23 (free acid basis)
Beilstein:
1911590
UNSPSC Code:
12352106
NACRES:
NA.25

Assay

≥95.0% (TLC)

storage temp.

−20°C

InChI

1S/C10H19O3P/c1-9(2)5-4-6-10(3)7-8-13-14(11)12/h5,7,14H,4,6,8H2,1-3H3,(H,11,12)/p-1

InChI key

KMVHUYNPESFWSO-UHFFFAOYSA-M

Application

Geranyl monophosphate (GP) may be used to study the kinetics and substrate specificity of modified/mutated isopentenyl phosphate kinases. Such studies may lead to engineered IPK mutants capable of facilitating the chemoenzymatic synthesis of longer chain isoprenoid diphosphates from monophosphate precursors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mark F Mabanglo et al.
ACS chemical biology, 7(7), 1241-1246 (2012-04-27)
Isopentenyl phosphate kinase (IPK) catalyzes the ATP-dependent phosphorylation of isopentenyl phosphate (IP) to form isopentenyl diphosphate (IPP) during biosynthesis of isoprenoid metabolites in Archaea. The structure of IPK from the archeaon Thermoplasma acidophilum (THA) was recently reported and guided the
Mo Chen et al.
Biochemistry, 49(1), 207-217 (2009-11-26)
Archaea synthesize isopentenyl diphosphate (IPP) and dimethylallyl diphosphate (DMAPP), the essential building blocks of isoprenoid compounds, from mevalonate (MVA). However, an analysis of the genomes of several members of the Archaea failed to identify genes for the enzymes required to
Nikki Dellas et al.
ACS chemical biology, 5(6), 589-601 (2010-04-16)
The biosynthesis of isopentenyl diphosphate (IPP) from either the mevalonate (MVA) or the 1-deoxy-d-xylulose 5-phosphate (DXP) pathway provides the key metabolite for primary and secondary isoprenoid biosynthesis. Isoprenoid metabolism plays crucial roles in membrane stability, steroid biosynthesis, vitamin production, protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service