51415C
Minimum Essential Medium
with Earle′s Balanced Salts, with 25mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Pharma Manufacturing
Synonym(s):
Minimum Essential Medium Eagle, MEM
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
description
for research or for further manufacturing use
Quality Level
sterility
sterile-filtered
form
liquid
technique(s)
cell culture | mammalian: suitable
components
Earle’s salts (5% CO2): yes
HEPES: 25 mM
L-glutamine: no
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
supplement
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Wenxin Wu et al.
Virology, 396(2), 178-188 (2009-11-17)
We studied cytokine responses to influenza virus PR8 (H1N1) and Oklahoma/309/06 (OK/06, H3N2) in a novel human lung tissue model. Exposure of the model to influenza virus rapidly activated the mitogen-activated protein kinase signaling (MAPK) pathways ERK, p38 and JNK.
Robert Nason et al.
Journal of the Association for Research in Otolaryngology : JARO, 10(2), 151-160 (2009-01-16)
Osteoclasts are the only cells capable of carrying out bone resorption and therefore are responsible for the osteolysis seen in infectious diseases such as chronic otitis media and infected cholesteatoma. Pseudomonas aeruginosa is the most common organism isolated from these
Chia-Chyi Liu et al.
PloS one, 3(3), e1810-e1810 (2008-03-20)
This work investigated the replication kinetics of the four dengue virus serotypes (DEN-1 to DEN-4), including dengue virus type 4 (DEN-4) recovered from an infectious cDNA clone, in Vero cells and in MRC-5 cells grown on Cytodex 1 microcarriers. DEN-1
Petra G Hirrlinger et al.
PloS one, 6(1), e16106-e16106 (2011-02-02)
Glial cells such as retinal Müller glial cells are involved in potassium ion and water homeostasis of the neural tissue. In these cells, inwardly rectifying potassium (Kir) channels and aquaporin-4 water channels play an important role in the process of
Dirk M Reiter et al.
PLoS pathogens, 7(8), e1002166-e1002166 (2011-08-11)
Many viruses attach to target cells by binding to cell-surface glycans. To gain a better understanding of strategies used by viruses to engage carbohydrate receptors, we determined the crystal structures of reovirus attachment protein σ1 in complex with α-2,3-sialyllactose, α-2,6-sialyllactose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service