Skip to Content
Merck
All Photos(1)

Documents

5125

Sigma-Aldrich

CDH13 human

recombinant, expressed in E. coli, 0.5 mg protein/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.75

biological source

human

recombinant

expressed in E. coli

description

0.1 mg recombinant human CDH13 in 20 mM Tris-HCl buffer containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

Assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 100 μg

concentration

0.5 mg protein/mL

technique(s)

cell culture | mammalian: suitable

accession no.

NP_001248

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CDH13(1012)

Application

Coating a plate well (6 well plate) with this recombinant CDH13 protein in a neuronal cell specific medium at 1-10 mg / well allows for 1) human neuronal cell and vascular endothelial cell / receptor interaction studies or 2) human neuronal and vascular endothelial cell/receptor interaction studies in vitro.

Use this procedure as a guideline to determine optimal coating conditions for the culture system of choice.
1. Thaw CDH13 and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so that the volume added covers the surface evenly (1-10 μg/well, 6 well plate).
2. Add appropriate amount of diluted material to culture surface.
3. Incubate at room temperature for approximately 1.5 hours.
4. Aspirate remaining material.
5. Rinse plates carefully with water and avoid scratching bottom surface of plates.
6. Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained.

Sequence

MASMTGGQQMGRGHHHHHHGNLYFQGGEFELSIVVSPILIPENQRQPFPRDVGKVVDSDRPERSKFRLTGKGVDQEPKGIFRINENTGSVSVTRTLDREVIAVYQLFVETTDVNGKTLEGPVPLEVIVIDQNDNRPIFREGPYIGHVMEGSPTGTTVMRMTAFDADDPATDNALLRYNIRQQTPDKPSPNMFYIDPEKGDIVTVVSPALLDRETLENPKYELIIEAQDMAGLDVGLTGTATATIMIDDKNDHSPKFTKKEFQATVEEGAVGVIVNLTVEDKDDPTTGAWRAAYTIINGNPGQSFEIHTNPQTNEGMLSVVKPLDYEISAFHTLLIKVENEDPLVPDVSYGPSSTATVHITVLDVNEGPVFYPDPMMVTRQEDLSVGSVLLTVNATDPDSLQHQTIRYSVYKDPAGWLNINPINGTVDTTAVLDRESPFVDNSVYTALFLAIDSGNPPATGTGTLLITLEDVNDNAPFIYPTVAEVCDDAKNLSVVILGASDKDLHPNTDPFKFEIHKQAVPDKVWKISKINNTHALVSLLQNLNKANYNLPIMVTDSGKPPMTNITDLRVQVCSCRNSKVDCNAAG

Preparation Note

The full-length extracellular domain of the human CDH13 gene (139-693 aa) was constructed with 31 N-terminal T7/HIS-tag and expressed in E. coli as inclusion bodies. The final product was refolded using our unique “temperature shift inclusion body refolding” technology and chromatographically purified.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danila Ivanov et al.
Experimental cell research, 293(2), 207-218 (2004-01-20)
T-cadherin (T-cad), an unusual glycosylphosphatidylinositol (GPI)-anchored member of the cadherin family of cell adhesion molecules, is widely expressed in the cardiovascular system. The expression profile of T-cad within diseased (atherosclerotic and restenotic) vessels indicates some relationship between expression of T-cad
Alexandra V Andreeva et al.
Journal of cellular physiology, 223(1), 94-102 (2009-12-30)
T-cadherin is an atypical member of the cadherin family, which lacks the transmembrane and intracellular domains and is attached to the plasma membrane via a glycosylphosphatidylinositol anchor. Unlike canonical cadherins, it is believed to function primarily as a signaling molecule.
Cristina Vergara et al.
Acta neuropathologica, 137(3), 397-412 (2019-01-02)
Neuropathological analysis in Alzheimer's disease (AD) and experimental evidence in transgenic models overexpressing frontotemporal dementia with Parkinsonism linked to chromosome 17 (FTDP-17) mutant tau suggest that amyloid-β pathology enhances the development of tau pathology. In this work, we analyzed this
Sujita Khanal et al.
PLoS pathogens, 15(2), e1007442-e1007442 (2019-03-01)
Persistent expression of high-risk HPV oncogenes is necessary for the development of anogenital and oropharyngeal cancers. Here, we show that E6/E7 expressing cells are hypersensitive to DNA crosslinking agent cisplatin and have defects in repairing DNA interstrand crosslinks (ICL). Importantly
Ada Ndoja et al.
Cell, 182(5), 1156-1169 (2020-08-17)
Dysregulated microglia are intimately involved in neurodegeneration, including Alzheimer's disease (AD) pathogenesis, but the mechanisms controlling pathogenic microglial gene expression remain poorly understood. The transcription factor CCAAT/enhancer binding protein beta (c/EBPβ) regulates pro-inflammatory genes in microglia and is upregulated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service