Skip to Content
Merck
All Photos(1)

Key Documents

50649

Sigma-Aldrich

Glyoxal solution

BioReagent, for molecular biology, ~40% in H2O (~8.8 M)

Synonym(s):

Ethanedial, Oxalaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHCCHO
CAS Number:
Molecular Weight:
58.04
Beilstein:
1732463
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.52

grade

Molecular Biology
for molecular biology

product line

BioReagent

form

liquid

concentration

~40% in H2O (~8.8 M)

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

refractive index

n20/D 1.409

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤5 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤300 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

[H]C(=O)C([H])=O

InChI

1S/C2H2O2/c3-1-2-4/h1-2H

InChI key

LEQAOMBKQFMDFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Glyoxal is a nucleic acid denaturant. It acts as a specific probe for homopurine strands of DNA.

Application

Suitable for
  • nucleic acid denaturation
  • binding of DNA/RNA to nitrocellulose membrane

Other Notes

This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.
May precipitate on storage; redissolve at 50-60 °C.

Caution

May precipitate on storage; redissolve at 50-60 °C.

Principle

Glyoxal forms stable complex with guanine residues that results in denaturation of nucleic acids.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Vojtisková et al.
Journal of biomolecular structure & dynamics, 5(2), 283-296 (1987-10-01)
Plasmid pEJ4, which is a derivative of pUC19 containing an insert with 60-bp-long homopurine.homopyrimidine tract from sea urchin P. miliaris histone gene spacer, was studied by chemical probes of the DNA structure osmium tetroxide and glyoxal. The former probe reacts
Hybridization of denatured RNA transferred or dotted nitrocellulose paper.
P S Thomas
Methods in enzymology, 100, 255-266 (1983-01-01)
R N Skopp et al.
Analytical biochemistry, 169(1), 132-137 (1988-02-15)
This paper describes agarose gel electrophoresis and silver staining of denatured RNAs. Glyoxal- or formaldehyde-denatured RNAs are electrophoresed in an agarose gel cast on a plastic support using an inert, low conductivity buffer. Following electrophoresis, the gel is stained with
Prasanna Channathodiyil et al.
PloS one, 16(1), e0240769-e0240769 (2021-01-23)
A simple method for extraction of high quality RNA from cells that have been fixed, stained and sorted by flow cytometry would allow routine transcriptome analysis of highly purified cell populations and single cells. However, formaldehyde fixation impairs RNA extraction
Catherine Lambert et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 18(1), 59-69 (2012-10-30)
The molecular basis of Alzheimer's disease has not been clearly established, but disruption of brain metal ion homeostasis, particularly copper and zinc, might be closely involved in the pathogenesis of this disease and its characteristic β-amyloid neuropathological features. The use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service