Skip to Content
Merck
All Photos(1)

Key Documents

49291

Sigma-Aldrich

Glucosidase from Aspergillus niger

powder, ≥750 U/g

Synonym(s):

Cellobiase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

powder

Quality Level

specific activity

≥750 U/g

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Glucosidase catalyzes the hydrolysis of α-1,4 linkages with a substrate preference for maltose, maltotriose and maltotetraose. Reactivity with large polysaccharides like dextrin and starch have also been described.
Glucosidase catalyzes the hydrolysis of α-1,4 linkages with a substrate preference for maltose, maltotriose and maltotetraose.[1][2] Reactivity with large polysaccharides like dextrin and starch have also been described.[3]

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol p-nitrophenyl-β-D-glucopyranoside per minute at pH 4.0 and 37 °C; may contain α- and β-glucosidase

Other Notes

Characterization[4][5]; Ethanol production from paper-mill waste fibre[6]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Action of α-D-glucosidase from Aspergillus niger towards dextrin and starch
M. Ota et al
Carbohydrate Polymers, 78, 287-291 (2009)
Substrate specificity and subsite affinities of crystalline α-glucosidase from Aspergillus niger
A. Kita et al
Agricultural and Biological Chemistry, 55, 2327-2335 (1991)
T Watanabe et al.
European journal of biochemistry, 209(2), 651-659 (1992-10-15)
Beta-glucosidase was purified from a crude cellulase preparation from Aspergillus niger by affinity chromatography on a methacrylamide-N-methylene-bis-methacrylamide copolymer bearing cellobiamine. The purified enzyme was a dimer with an isoelectric point of 4.0. The molecular mass of the enzyme was estimated
McCleary, B.V., and Harrington, J.,
Methods in Enzymology, 160, 575-575 (1988)
Xiaohong Lan et al.
Food chemistry, 188, 632-640 (2015-06-05)
Starches from five underutilized tubers (canna, potato, Chinese yam, water chestnut, and taro) were extracted to investigate quantitative structure-property relationships (QSPR) in each starch using a combination of X-ray diffraction (XRD) and small-angle X-ray scattering (SAXS). Structural parameters of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service