Skip to Content
Merck
All Photos(1)

Documents

487R-9

SMAD4 (MRQ-72) Rabbit Monoclonal Antibody

For In Vitro Diagnostic Use in Select Regions

Synonym(s):

Mothers Against Decapentaplegic Homolog 4 Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

MRQ-72, monoclonal

packaging

vial of 0.1 mL concentrate (487R-94)
vial of 0.1 mL concentrate (487R-94-RUO) Research Use Only
vial of 0.5 mL concentrate (487R-(95)
vial of 1.0 mL (concentrate (487R-96))
vial of 1.0 mL concentrate (487R-96-RUO) Research Use Only
vial of 1.0 mL pre-dilute (487R-97-RUO) Research Use Only
vial of 1.0 mL pre-dilute ready-to-use (487R-97)
vial of 7.0 mL pre-dilute ready-to-use (487R-98)
vial of 7.0 mL pre-dilute ready-to-use (487R-98-RUO) Research Use Only

manufacturer/tradename

Cell Marque

isotype

IgG

control

pancreas

visualization

cytoplasmic, nuclear

General description

Mothers Against Decapentaplegic Homolog 4 (SMAD4) is a transcription factor that is involved in TGFβ signalling pathways and acts as a tumor suppressor. SMAD4 is commonly expressed in a variety of cancers, including pancreatic ductal adenocarcinoma (PDA), colorectal carcinoma (CRC), hepatocellular carcinoma (HCC), and gastric carcinomas, as well as non-neoplastic liver, pancreas, and colon. However, a loss of expression has been observed in a subset of PDA, CRC, and gastric carcinomas due to a variety of mutations including nonsense, missense, deletions, and splice site changes. In contrast, SMAD4 is over-expressed in HCC compared to the weak expression that is exhibited in the non-neoplastic liver.

Quality

IVD - MQ500

RUO - MQ100

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format

Note: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Torbenson et al.
Human pathology, 33(9), 871-876 (2002-10-16)
In the normal liver, the transforming growth factor beta (TGF-beta) signaling pathway plays an important role in inhibiting hepatocyte growth. This effect is mediated through Smad4 (or Dpc4), a tumor-suppressor gene that affects gene transcription and controls cell growth. A
K L Woodford-Richens et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(17), 9719-9723 (2001-08-02)
Loss of chromosome 18q21 is well documented in colorectal cancer, and it has been suggested that this loss targets the DCC, DPC4/SMAD4, and SMAD2 genes. Recently, the importance of SMAD4, a downstream regulator in the TGF-beta signaling pathway, in colorectal
Y H Kim et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 15(4), 574-580 (2004-03-23)
Transforming growth factor-beta (TGF-beta) modulates the growth and function of many cells, including those with malignant transformation. Smad proteins have been identified as major components in the intracellular signaling of TGF-beta family members. To clarify the correlations between clinicopathologic profiles
R Salovaara et al.
Gut, 51(1), 56-59 (2002-06-22)
Loss of DNA sequences from chromosome 18q21 is a major genetic change in colorectal tumorigenesis. Multiple genes have been identified in this area. One of these, DPC4 (deleted in pancreatic cancer 4, also known as SMAD4), is mutated in a
Lauren L Ritterhouse et al.
Histopathology, 75(4), 546-551 (2019-05-06)
SMAD4 (DPC4) is a tumour suppressor gene that is dysregulated in various tumour types, particularly pancreaticobiliary and gastrointestinal carcinomas. Corresponding loss of protein expression has been reported in approximately 50% of pancreatic and 25% of colonic adenocarcinomas. In the evaluation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service