Skip to Content
Merck
All Photos(1)

Key Documents

453R-1

Sigma-Aldrich

p53 (SP5) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP5, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (453R-14)
vial of 0.5 mL concentrate (453R-15)
bottle of 1.0 mL predilute (453R-17)
vial of 1.0 mL concentrate (453R-16)
bottle of 7.0 mL predilute (453R-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

colon carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... TP53(7157)

General description

Anti-p53 tumor suppressor protein antibody recognizes a 53 kDa phosphoprotein, identified as p53 suppressor gene product. It reacts with the mutant as well as wild type p53, although significant accumulation of the mutant form of p53 protein due to longer half-life half-life is the basis for the test using the IHC technique. Quantitation of nuclear staining with this antibody has been shown to be a factor in breast carcinoma, lung carcinoma, colorectalcarcinoma, urothelial carcinoma, and ependymoma. p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia.

Linkage

p53 Positive Control Slides, Product No. 453S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diagnostic Immunohistochemistry: Theranostic and Genomic Applications (2006)
A Korshunov et al.
Journal of the neurological sciences, 177(1), 72-82 (2000-09-01)
Intracranial ependymomas are the third most common primary brain tumor in children. Although clinical and histological criteria for ependymoma prognosis are recognized, studies have reported contradictory results. Prognostic significance based on immunohistochemistry of ependymomas has been reported in a few
B E Moore et al.
Applied immunohistochemistry & molecular morphology : AIMM, 9(3), 203-206 (2001-09-15)
Intratubular germ cell neoplasia, unclassified (IGCNU) is a precursor of germ cell tumors (GCT) of the testis. In routine histologic sections, neoplastic intratubular germ cells may be very few and easily overlooked. The aim of this study is two-fold: to
D C Quinlan et al.
Cancer research, 52(17), 4828-4831 (1992-09-01)
Mutations in the gene coding for the p53 tumor suppressor protein are common in a variety of human cancers. To assess the role of a putative mutated p53 protein in human lung cancer, a monoclonal antibody recognizing it was used
C Midulla et al.
Anticancer research, 19(5B), 4033-4037 (2000-01-11)
The different clinical evolution of breast cancer with similar pathological characteristics prompted the authors to investigate the prognostic significance of different biological markers. Seventy-one primary breast carcinoma specimens obtained by mastectomy or quadrantectomy were examined for the determination of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service