Skip to Content
Merck
All Photos(2)

Key Documents

450R-1

Sigma-Aldrich

Prostein (EP381) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP381, monoclonal

description

This product is intended for research use only. Not for use in diagnostic procedures.

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate Research Use Only (450R-14-RUO)
vial of 1.0 mL concentrate Research Use Only (450R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (450R-17-RUO)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (450R-18-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100 (concentrated)

isotype

IgG

control

prostate, prostate carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Prostein is a type IIIA transmembrane protein comprising a cleavable signal peptide and 11 transmembrane domains. In vitro studies demonstrate that prostein transcription and expression is androgen responsive. Prostein expression was found to be restricted to tissues of prostatic origin such as non-neoplastic prostate, prostate lesions, and malignant prostate tissues. Due to its high specificity, immunohistochemical detection of prostein may have utility in distinguishing prostatic carcinoma from the carcinomas of bladder, kidney, lung and colorectum.

Quality


IVD

IVD

IVD

RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd Sheridan et al.
The American journal of surgical pathology, 31(9), 1351-1355 (2007-08-28)
Adenocarcinoma of the prostate can present as metastatic carcinoma with no known primary. Prostatic origin can be confirmed in most of these cases by immunohistochemistry for prostate-specific antigen (PSA) and prostate-specific acid phosphatase. In a small subset of high-grade prostate
Michael Kalos et al.
The Prostate, 60(3), 246-256 (2004-06-04)
Prostein is a recently described molecule expressed at the mRNA level in a prostate-specific manner. A murine monoclonal antibody was developed, characterized, and used to evaluate the expression of prostein protein in prostatic, other normal tissue and tumor samples. The
J Xu et al.
Cancer research, 61(4), 1563-1568 (2001-03-14)
In this report, we describe the application of a systematic, genome-based approach to identify prostein, a novel prostate-specific protein expressed in normal and malignant prostate tissues. Characterization of the prostein gene shows that prostein cDNA encodes a 553-amino acid protein.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service