Skip to Content
Merck
All Photos(1)

Key Documents

312R-2

Sigma-Aldrich

PAX-5 (EP156) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP156, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (312R-24)
vial of 0.5 mL concentrate (312R-25)
bottle of 1.0 mL predilute (312R-27)
vial of 1.0 mL concentrate (312R-26)
bottle of 7.0 mL predilute (312R-28)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... PAX5(5079)

General description

PAX-5 encodes for B-cell-specific activator protein (BSAP), a marker for B-cells, including B-lymphoblastic neoplasms and maturation stage. It is found in most cases of mature and precursor B-cell non-Hodgkin lymphomas/leukemias. In approximately 97% of cases of classic Hodgkin lymphoma, Reed-Sternberg cells express PAX-5. PAX-5 is not detected in multiple myeloma and solitary plasmacytoma, making it useful for such differentiation. Diffuse large B-cell lymphomas do express PAX-5, save for those with terminal B-cell differentiation. T-cell neoplasms do not stain with anti-PAX-5. There is a strong association with CD20 expression.

Quality


IVD

IVD

IVD

RUO

Linkage

PAX-5 Positive Control Slides, Product No. 312S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brunangelo Falini et al.
Blood, 99(2), 409-426 (2002-01-10)
Acquired chromosomal anomalies (most commonly translocations) in lymphoma and leukemia usually result in either activation of a quiescent gene (by means of immunoglobulin or T-cell-receptor promotors) and expression of an intact protein product, or creation of a fusion gene encoding
Ines Schwering et al.
Blood, 101(4), 1505-1512 (2002-10-24)
Hodgkin and Reed-Sternberg (HRS) cells represent the malignant cells in classical Hodgkin lymphoma (HL). Because their immunophenotype cannot be attributed to any normal cell of the hematopoietic lineage, the origin of HRS cells has been controversially discussed, but molecular studies
Emina Torlakovic et al.
The American journal of surgical pathology, 26(10), 1343-1350 (2002-10-03)
Whereas L26 (anti-CD20) is well established as a B-cell marker of high specificity for use in paraffin-embedded tissues and JCB117 (anti-CD79a) is increasingly used, a comparable additional pan-B-cell antibody has hitherto not yet been identified. Here we have studied the
Klaus Willenbrock et al.
Laboratory investigation; a journal of technical methods and pathology, 82(9), 1103-1109 (2002-09-10)
The atypical cells of CD30(+) cutaneous lymphoproliferative disorders (CD30CLD) are commonly of T-cell origin and frequently have a similar morphology as Hodgkin or Reed-Sternberg cells of Hodgkin's lymphoma (HL). HL is one of the tumors associated with CD30CLD. Although most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service