Skip to Content
Merck
All Photos(1)

Key Documents

303M-1

Sigma-Aldrich

Cytokeratin, HMW (AE3) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AE3, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (303M-14)
vial of 0.5 mL concentrate (303M-15)
bottle of 1.0 mL predilute (303M-17)
vial of 1.0 mL concentrate (303M-16)
bottle of 7.0 mL predilute (303M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

prostate

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... KRT1(3848)

General description

Anti-cytokeratin, high molecular weight (AE3) is capable of recognizing all basic keratins; therefore, it is a broadly reactive antibody staining most epithelia and their neoplasms. Members of the acidic and basic subfamilies are found together in pairs. Since each epithelium contains at least one acidic and one basic keratin, this antibody is used to observe the distribution of keratin-containing cells in normal epithelia and to identify neoplasms derived from such epithelium.

Quality


IVD

IVD

IVD

RUO

Linkage

Cytokeratin, HMW Positive Control Slides, Product No. 303S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoclonal antibody studies of mammalian epithelial keratins: a review.
T T Sun et al.
Annals of the New York Academy of Sciences, 455, 307-329 (1985-01-01)
W G Nelson et al.
Cancer research, 44(4), 1600-1603 (1984-04-01)
The expression of specific keratin polypeptides in human neoplasms was investigated by the immunoblot technique using monoclonal anti-keratin antibodies. Mr 50,000 and 58,000 keratins, recognized by AE1 and AE3 antibodies, respectively, were detected only in carcinomas of stratified epithelial origin
P E Swanson
American journal of clinical pathology, 95(4 Suppl 1), S2-S7 (1991-04-01)
Historically, antibodies to cytokeratin intermediate filaments have been models of target specificity. In most diagnostic settings, the utility of these antibodies was unquestioned; reactivity for cytokeratin was dogmatically equated with epithelial differentiation. Recently, however, the diagnostic importance of these antibodies
V Eusebi et al.
The American journal of surgical pathology, 14(8), 737-747 (1990-08-01)
We present four cases of a malignant thyroid tumor showing morphologic, immunocytochemical, and ultrastructural features of endothelial cell differentiation. The tumor cells had epithelioid features and displayed strong immunoreactivity for keratin. There was no evidence of follicular or C-cell differentiation
A Lopez-Beltrán et al.
Virchows Archiv : an international journal of pathology, 438(6), 552-557 (2001-07-27)
Lymphoepithelioma-like carcinoma (LELCA) of the urinary bladder is a rare variant of bladder cancer characterized by a malignant epithelial component densely infiltrated by lymphoid cells. It is characterized by indistinct cytoplasmic borders and a syncytial growth pattern. These neoplasms deserve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service