Skip to Content
Merck
All Photos(1)

Documents

302M-1

Sigma-Aldrich

Neurofilament (2F11) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

2F11, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (302M-14)
vial of 0.5 mL concentrate (302M-15)
bottle of 1.0 mL predilute (302M-17)
vial of 1.0 mL concentrate (302M-16)
bottle of 7.0 mL predilute (302M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

brain

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... NEFL(4747)

General description

Anti-neurofilament stains an antigen localized in a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament.

Quality


IVD

IVD

IVD

RUO

Linkage

Neurofilament Positive Control Slides, Product No. 302S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Dubovy et al.
The British journal of ophthalmology, 85(8), 949-951 (2001-07-24)
Palisaded encapsulated neuroma of the skin (PEN) is an uncommon benign neural tumour that occurs on the head and neck, including the eyelid, but is unfamiliar to many ophthalmic clinicians. For the first time in the ophthalmic literature, the clinical
Protease inhibitors of human plasma. Alpha 1-antichymotrypsin.
R W Berninger
Journal of medicine, 16(1-3), 101-128 (1985-01-01)
J N Wood et al.
Bioscience reports, 1(3), 263-268 (1981-03-01)
Hybrid cell lines secreting monoclonal antibodies against mammalian neurofilaments have been prepared. An improved protocol for the preparation of neurofilaments and methods for the identification and isolation of such hybridomas are presented. The antibodies produced specifically stain only neuronal
Irma E Erana-Rojas et al.
Annals of diagnostic pathology, 6(5), 265-271 (2002-10-12)
Demyelinating disease presenting as a solitary contrast-enhancing mass poses a diagnostic challenge for both radiologists and surgical pathologists. We report the cases of two female patients, aged 23 and 37 years, who exhibited the clinical and radiologic features of a
G N van Muijen et al.
The American journal of pathology, 116(3), 363-369 (1984-09-01)
Three monoclonal antibodies, one directed against cytokeratin (clone 80) and two directed against neurofilament (clones 2F11 and 3G6), were used in the study of a series of 77 lung carcinomas by immunohistochemical staining. The anti-cytokeratin antibody, a very broadly reacting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service