Skip to Content
Merck
All Photos(1)

Key Documents

25810

Sigma-Aldrich

Chloroperoxidase from Caldariomyces fumago

aqueous suspension, brown, >10,000 U/mL

Synonym(s):

Chloride Peroxidase, Chloride:hydrogen-peroxide oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fungus (Caldariomyces fumago)

Quality Level

form

aqueous suspension

concentration

>10,000 U/mL

color

brown

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Chloroperoxidase (CPO) is a major heme-containing, synthetic and versatile enzyme, obtained from Caldariomyces fumago.

Application

A useful alternative to lactoperoxidase for 131I ion labeling studies, for bromination of proteins, and for 36Cl labeling of macromolecules in long-term isolation procedures.

Biochem/physiol Actions

Chloroperoxidase (CPO) helps in the catalysis of oxidation reactions with the help of hydrogen peroxide. It displays peroxidase, catalase and cytochrome P450-like functions. CPO also plays a role in catalyzing halogenation reactions.

Unit Definition

1 U corresponds to the amount of enzyme which converts 1 μmol of monochlorodimedone to dichlorodimedone per minute at pH 2.75 and 25 °C in the presence of KCl and H2O2.

Physical form

Supplied as a suspension in 0.1 M sodium phosphate pH 4.0.

Other Notes

Oxidation of aminopyrine; Chloroperoxidase, a peroxidase with potential; Chloroperoxidase-catalyzed asymmetric transformations.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Sayo et al.
Chemical & pharmaceutical bulletin, 37(12), 3347-3350 (1989-12-01)
Although in the absence of halide ion chloroperoxidase did not catalyze the ethylhydroperoxide (EHP)-supported oxidation of aminopyrine, in the presence of Br- or Cl-, chloroperoxidase did catalyze the oxidation of aminopyrine, generating the aminopyrine cation radical (AP+). The initial rate
H. Fu et al.
The Journal of Organic Chemistry, 57, 7265-7265 (1992)
M.A. Pickard et al.
Journal of Industrial Microbiology, 7, 235-235 (1991)
Fast and efficient purification of chloroperoxidase from C. fumago
Yazbik V and Ansorge SM
Process. Biochem., 45(2), 279-283 (2010)
S. Colonna et al.
NATO ASI Ser., Ser. C, 381, 323-323 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service