Skip to Content
Merck
All Photos(1)

Key Documents

231R-1

Sigma-Aldrich

Calponin-1 (EP798Y) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP798Y, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (231R-14)
vial of 0.5 mL concentrate (231R-15)
bottle of 1.0 mL predilute (231R-17)
vial of 1.0 mL concentrate (231R-16)
bottle of 7.0 mL predilute (231R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200

isotype

IgG

control

appendix

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Calponin is a 34-kD polypeptide that interacts with actin, tropomyosin, and calmodulin. Itis involved in the smooth muscle contraction mechanism and is restricted exclusively to smoothmuscle tissue. Anti-calponin has been found to be useful in staining myoepithelium and thushas been used to confirm the presence of myoepithelial cells in benign breast proliferationand their absence in invasive breast carcinomas. Calponin positivity has also been noted inmyoepithelial neoplasms and pleomorphic adenoma of the salivary gland.

Quality


IVD

IVD

IVD

RUO

Linkage

Calponin Positive Control Slides, Product No. 231S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheryl B Hill et al.
American journal of clinical pathology, 123(1), 36-44 (2005-03-15)
We evaluated 25 intraductal papillomas and 18 papillary carcinomas (invasive, 4; micropapillary ductal carcinoma in situ [DCIS], 5; cases originally classified as intracystic/intraductal papillary carcinoma, 9) by calponin, smooth muscle myosin heavy chain (SMM-HC), and p63 immunostains. Calponin, SMM-HC, and
Antibodies to novel myoepithelium-associated proteins distinguish benign lesions and carcinoma in situ from invasive carcinoma of the breast
Wang NP, et al.
Appl. Immunohistochem., 5, 141-151 (1997)
A T Savera et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 10(11), 1093-1100 (1997-12-05)
Myoepithelial cells of salivary glands have a complex cytoskeletal immunophenotype. To elaborate the smooth muscle phenotype of salivary gland myoepithelium and to assess its contribution to the histogenesis of pleomorphic adenomas, we evaluated the immunohistochemical expression of three novel monoclonal
J C Fanburg-Smith et al.
Human pathology, 30(11), 1336-1343 (1999-11-26)
Angiomatoid "malignant" fibrous histiocytoma (AMFH) has been considered to be a low-grade sarcoma of childhood, and, with its fibrous pseudocapsule, angiomatoid change, dense lymphoplasmacytic response, and proliferation of spindled or round cells, has been classified as a fibrohistiocytic neoplasm. We
Jason L Hornick et al.
The American journal of surgical pathology, 27(9), 1183-1196 (2003-09-10)
Myoepitheliomas and mixed tumors were only recently recognized to occur primarily in soft tissue, and only small case numbers have been described. To characterize these tumors further and to evaluate prognostic parameters, 101 myoepithelial tumors of soft tissue were retrieved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service