Skip to Content
Merck
All Photos(1)

Key Documents

C85700

Sigma-Aldrich

o-Cresol

ReagentPlus®, ≥99%

Synonym(s):

2-Methylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Beilstein:
506917
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent
bp:
191 °C (lit.)
vapor pressure:
0.3 mmHg ( 20 °C)

grade

reagent

vapor density

3.72 (vs air)

vapor pressure

0.3 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

solid or liquid

autoignition temp.

1110 °F

expl. lim.

1.47 %, 148 °F

dilution

(for general lab use)

pH

4.5 (25 g/L)

bp

191 °C (lit.)

mp

29-31 °C (lit.)

SMILES string

Cc1ccccc1O

InChI

1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

QWVGKYWNOKOFNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna M Dela Cruz et al.
Journal of mass spectrometry : JMS, 42(2), 178-186 (2006-12-19)
The influence shaped femtosecond laser pulses have on molecular photofragmentation and ionization, coupled with the intrinsic sensitivity of mass spectrometry, results in a powerful tool for fast, accurate, reproducible and quantitative isomeric identification. Complex phase functions are introduced to enhance
Irama Sanchez et al.
Chemosphere, 68(2), 338-344 (2007-02-16)
This work deals with a new abiotic oxidation process designed as a suitable pre-treatment step within a biological depuration of wastewater containing phenol or its derivatives (o-cresol, 2-chlorophenol and p-nitrophenol) or aniline. The reaction was carried out in a stirred
Hua-Wei Chen et al.
Water research, 41(10), 2069-2078 (2007-04-10)
Platinum deposited TiO(2) films were prepared on quartz substrates by dip-coating process for the photodecomposition of o-cresol. The characteristics of Pt/TiO(2) and the temporal behavior of o-cresol decomposition by Pt/TiO(2) photocatalysis under visible light irradiation were investigated. Platinum deposited on
Hua-Wei Chen et al.
Chemosphere, 69(2), 184-190 (2007-06-05)
This study investigated the effect of controlled periodic illumination on the photonic efficiency and temporal decomposition behavior of o-cresol by UV/TiO2 process in a slurry reactor using light emitting diodes as the light source. Based on the same UV light
María Eugenia Suarez-Ojeda et al.
Chemosphere, 66(11), 2096-2105 (2006-11-11)
This study examines the feasibility of coupling a Catalytic Wet Air Oxidation (CWAO), with activated carbon (AC) as catalyst, and an aerobic biological treatment to treat a high-strength o-cresol wastewater. Two goals are pursued: (a) To determine the effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service