Skip to Content
Merck
All Photos(1)

Key Documents

91707

Sigma-Aldrich

Trifluoroacetic acid

Trifluoroacetic acid

puriss. p.a., suitable for HPLC, ≥99.0% (GC)

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable

impurities

≤0.05% water

refractive index

n20/D 1.3 (lit.)

pH

1 (20 °C, 1 g/L)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

solubility

water: soluble

density

1.489 g/mL at 20 °C (lit.)

λ

1 cm path, H2O reference

UV absorption

λ: 260 nm Amax: 0.9
λ: 270 nm Amax: 0.1
λ: 280 nm Amax: 0.08
λ: 290 nm Amax: 0.07
λ: 300 nm Amax: 0.06
λ: 320 nm Amax: 0.05

suitability

corresponds to standard for RP gradient test

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Trifluoroacetic acid (TFA) is a fluorinated acetic acid. It is widely used as trifluoromethylating reagent for trifluoromethylation of aromatic compounds.[1]

Application

Trifluoroacetic acid (TFA) may be used in the following studies:
  • Preparation of TFA reagent for the estimation of serum vitamin A.[2]
  • To compose the binary solvent system for the isolation of large denatured peptides by reverse phase high performance liquid chromatography.[3]
  • A mobile-phase additive for liquid chromatography-mass spectrometry (LC-MS) analysis.[4]
  • Preparation of artemisinin under optimized reaction conditions.[5]

Packaging

package with 10x1mL

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Continuous-flow synthesis of the anti-malaria drug artemisinin.
François Lévesque et al.
Angewandte Chemie (International ed. in English), 51(7), 1706-1709 (2012-01-18)
Macro- and micromethods for the determination of serum vitamin A using trifluoroacetic acid.
J B Neeld et al.
The Journal of nutrition, 79, 454-462 (1963-04-01)
Guangfa Shi et al.
Organic letters, 17(1), 38-41 (2014-12-18)
Direct trifluoromethylation of arenes using TFA as the trifluoromethylating reagent was achieved with Ag as the catalyst. This reaction not only provides a new protocol for aryl C-H trifluoromethylation, but the generation of CF3· from TFA may prove useful in
Risto Kostiainen et al.
Journal of chromatography. A, 1216(4), 685-699 (2008-09-19)
The most widely used ionization techniques in liquid chromatography-mass spectrometry (LC-MS) are electrospray ionization (ESI), atmospheric pressure chemical ionization (APCI) and atmospheric pressure photoionization (APPI). All three provide user friendly coupling of LC to MS. Achieving optimal LC-MS conditions is
W C Mahoney et al.
The Journal of biological chemistry, 255(23), 11199-11203 (1980-12-10)
A method employing reverse phase high performance liquid chromatography has been developed for the purification of large denatured peptides. The cyanogen bromide fragments of hemoglobin alpha, beta, and gamma chains (13 to 91 residues in length) could be separated and

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service