Skip to Content
Merck
All Photos(3)

Key Documents

72253

Sigma-Aldrich

Nickel(II) nitrate hexahydrate

purum p.a., crystallized, ≥97.0% (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
290.79
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystals

grade

purum p.a.

Assay

≥97.0% (KT)

form

crystals

quality

crystallized

mp

56 °C (lit.)

density

2.05 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤500 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤100 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Ni.6H2O/c2*2-1(3)4;;;;;;;/h;;;6*1H2/q2*-1;+2;;;;;;

InChI key

AOPCKOPZYFFEDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nickel(II) nitrate hexahydrate (Ni(NO3)2.6H2O) is a hydrated nickel(II) salt. Its thermal degradation has been investigated by thermogravimetric methods under quasi-isothermal conditions.

Application

Nickel(II) nitrate hexahydrate may be used as precursor for the synthesis of [Ni1/3Co1/3Mn1/3]O2 powders and Ni (II) oxide aerogels.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 2 - Ox. Sol. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal decomposition of nickel nitrate hexahydrate, Ni(NO3)2?6H2O, in comparison to Co(NO3)2?6H2O and Ca(NO3)2?4H2O.
Brockner W, et al.
Thermochimica Acta, 456(1), 64-68 (2007)
Monolithic nickel (II)-based aerogels using an organic epoxide: the importance of the counterion.
Gash AE, et al.
Journal of Non-Crystalline Solids, 350, 145-151 (2004)
H Tanojo et al.
Acta dermato-venereologica. Supplementum, (212)(212), 19-23 (2002-01-23)
Allergic contact dermatitis due to nickel salts is common. It is therefore important to measure the permeation of these salts through the stratum corneum (SC), the primary rate-limiting domain in skin. An advanced diffusion system and analytical techniques now enable
Jun Li et al.
Journal of colloid and interface science, 332(1), 231-236 (2009-01-16)
In this paper, we report the successful synthesis of Ni(12)P(5) hollow spheres via a facile hydrothermal route, employing white phosphorus (WP) and nickel nitrate as the reactants in the presence of hexamethylenetetramine (HMT) and polyethylene glycol 10000 (PEG-10000). The phase
Natalie Baho et al.
Inorganic chemistry, 46(1), 299-308 (2007-01-03)
The complexes [(dpdpm)Ni(2-NO3)2] (1), [(dpdpm)Ni(2-NO3)(1-NO3)(CH3CN)] (2), [(dpdpm)2Ni(1-NO3)(H2O)]NO3 (3), and [(dpdpm)2Ni(H2O)2][NO3]2 (4) (dpdpm = diphenyl(dipyrazolyl)methane, Ph2C(C3N2H3)2), have been prepared and characterized by IR and UV-vis-NIR spectroscopy and X-ray diffraction studies. X-ray studies have confirmed that complexes 1-4 all adopt variously distorted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service