Skip to Content
Merck
All Photos(3)

Key Documents

702587

Sigma-Aldrich

Potassium ferricyanide(III)

powder or chunks, <10 μm, 99%

Synonym(s):

Potassium hexacyanoferrate(III), Red prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K3Fe(CN)6
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
99%
form:
powder or chunks
solubility:
water: soluble 329 g/L at 20 °C

Quality Level

Assay

99%

form

powder or chunks

particle size

<10 μm

pH

6-9 (25 °C, 329 g/L)

solubility

water: soluble 329 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%

SMILES string

[K+].[K+].[K+].N#C[Fe-3](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.3K/c6*1-2;;;;/q;;;;;;-3;3*+1

InChI key

MIMJFNVDBPUTPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium ferricyanide(III) is a potassium salt widely employed as an external indicator in potassium dichromate titrations. A sensor containing potassium ferricyanide assisted enzyme electrode for estimating maltose and glucose in samples has been reported. Addition of potassium ferricyanide in the catholyte can improve the generation of power in microbial fuel cells (MFCs).

Application

Potassium ferricyanide(III) has been used:
  • To prepare the X-gal staining solution for staining cryosectioned tissues of injured neonatal mouse heart.
  • Reducing power assay of freeze-dried ethanolic extract of Issopus officinalis.
  • In combination with glutaraldehyde and osmium tetroxide for post-fixation of biological samples prior to epoxy embedding.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium ferricyanide mediated disposable biosensor for determination of maltose and glucose.
Ge F, et al.
Analytical Letters, 31(3), 383-394 (1998)
The obtaining of an antioxidant product based on Issopus officinalis freeze: Dried extract.
Emanuel V, et al.
Journal of Medicinal Plants Research, 5(10), 2078-2085 (2011)
Thomas C Jhou et al.
Neuron, 61(5), 786-800 (2009-03-17)
Separate studies have implicated the lateral habenula (LHb) or amygdala-related regions in processing aversive stimuli, but their relationships to each other and to appetitive motivational systems are poorly understood. We show that neurons in the recently identified GABAergic rostromedial tegmental
Thomas Stone et al.
Current biology : CB, 27(20), 3069-3085 (2017-10-11)
Path integration is a widespread navigational strategy in which directional changes and distance covered are continuously integrated on an outward journey, enabling a straight-line return to home. Bees use vision for this task-a celestial-cue-based visual compass and an optic-flow-based visual
Effects of cathodic electron acceptors and potassium ferricyanide concentrations on the performance of microbial fuel cell.
Wei L, et al.
International Journal of Hydrogen Energy, 37(17), 12980-12986 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service