Skip to Content
Merck
All Photos(1)

Key Documents

673811

Sigma-Aldrich

Diethyl ether

Diethyl ether

≥99.0%, anhydrous, ACS reagent, contains BHT as inhibitor

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.07
grade:
ACS reagent
anhydrous
bp:
34.6 °C (lit.)
vapor pressure:
28.66 psi ( 55 °C)
8.56 psi ( 20 °C)
Pricing and availability is not currently available.

Product Name

Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor

grade

ACS reagent
anhydrous

Quality Level

vapor density

2.6 (vs air)

vapor pressure

28.66 psi ( 55 °C)
8.56 psi ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

320 °F

contains

BHT as inhibitor

expl. lim.

36.5 %

dilution

(for analytical testing)

impurities

Alcohol (as CH3CH2OH), passes test
≤0.0002 meq/g Titr. acid
≤0.001% Carbonyl (as HCHO)
≤0.03% Water
≤1 ppm Peroxide (as H2O2)

evapn. residue

≤0.001%

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl ether (DEE) is a flammable and highly volatile organic compound commonly used as a solvent in organic synthesis. DEE can generate organic peroxides when stored for a long time, which can be prevented by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT scavenges the free radicals required for peroxide formation.

Application

Diethyl ether ACS grade solvent meets or exceeds the high standards of the American Chemical Society (ACS). It is ideal for most research needs and delivers replicable, publishable results.
Diethyl ether ACS grade solvent is used in:      
  • Routine chemical synthesis      
  • Drying & purification      
  • Critical labware cleaning

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Miriam Oberholzer et al.
Journal of visualized experiments : JoVE, (85)(85), doi:10-doi:10 (2014-04-02)
Dichloro-bis(aminophosphine) complexes of palladium with the general formula of [(P{(NC5H10)3-n(C6H11)n})2Pd(Cl)2] (where n = 0-2), belong to a new family of easy accessible, very cheap, and air stable, but highly active and universally applicable C-C cross-coupling catalysts with an excellent functional
Radical steps in diethyl ether decomposition.
Seres I and Huhn P.
International Journal of Chemical Kinetics, 18(8), 829-836 (1986)
Metabolism of diethyl ether and cometabolism of methyl tert-butyl ether by a filamentous fungus, a Graphium sp.
Hardison LK, et al.
Applied and Environmental Microbiology, 63(8), 3059-3067 (1997)
Vacuum Ultraviolet Absorption Spectrum of Diethyl Ether.
Hernandez GJ.
J. Chem. Phys. , 39(5), 1355-1355 (1963)
Jenny Liu et al.
Journal of visualized experiments : JoVE, (80)(80), doi:10-doi:10 (2013-10-23)
Convenient methods for the rapid, parallel synthesis of diversely functionalized nanoparticles will enable discovery of novel formulations for drug delivery, biological imaging, and supported catalysis. In this report, we demonstrate parallel synthesis of brush-arm star polymer (BASP) nanoparticles by the

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. Massachusetts
    • Reviews 9
    • Vote 1
    5 out of 5 stars.

    In stock, good value

    Arrived on time and correct product was delivered.

    Helpful?

  2. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service