Skip to Content
Merck
All Photos(1)

Key Documents

53847

Sigma-Aldrich

Hydrazine hydrate solution

puriss. p.a., 24-26% in H2O (RT)

Synonym(s):

Hydrazinium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2NH2 · H2O
CAS Number:
Molecular Weight:
50.06
Beilstein:
878137
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

form

liquid

concentration

24-26% (iodometric)
24-26% in H2O (RT)

density

1.01 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.NN

InChI

1S/H4N2.H2O/c1-2;/h1-2H2;1H2

InChI key

IKDUDTNKRLTJSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrazine hydrate solution is the 24-26% aqueous solution of hydrazine.

Application

Hydrazine hydrate solution (24-26% in H2O) is used as a reducing agent in chemical synthesis. It is used to prepare nickel(0) nanoparticles via in-situ reduction of NiCl2 salt.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G M Murphy et al.
Journal of clinical pathology, 23(7), 594-598 (1970-10-01)
The enzymatic technique using 3alpha-hydroxysteroid dehydrogenase for determining bile acids in blood has been modified by measuring the reduced nicotinamide adenine dinucleotide fluorimetrically. The increased sensitivity attained enables the concentration of total bile acids in serum to be estimated using
S D Turley et al.
Journal of lipid research, 19(7), 924-928 (1978-09-01)
A review of the 3 alpha-hydroxysteroid dehydrogenase method for determining the concentration of total bile acids in bile is described. The optimum conditions for the assay were established with respect to pH, temperature, incubation time, amount of NAD+, and units
Synthesis of Au, Ag and Au-Ag alloy nanoparticles in aqueous polymer solution.
Pal A, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 302(1), 51-57 (2007)
Yunpeng Huang et al.
Nanoscale, 6(18), 10673-10679 (2014-08-05)
In this work, we report the fabrication of low crystalline, few-layered MoS₂ nanosheet-coated electrospun SnO₂ nanotube (MoS₂/SnO₂) heterostructures with three-dimensional configurations by electrospinning combined with a one-step solvothermal approach. The morphologies and compositions of the as-prepared hybrid nanotubes were characterized
Yunpeng Huang et al.
Nanoscale, 7(44), 18595-18602 (2015-10-23)
Exploration of high-efficiency Pt-free electrochemical catalysts for hydrogen evolution reaction (HER) is considered as a great challenge for the development of sustainable and carbon dioxide free energy conversion systems. In this work, a unique hierarchical nanostructure of few-layered MoSe2 nanosheets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service