Skip to Content
Merck
All Photos(4)

Documents

402974

Sigma-Aldrich

Lithium hydroxide monohydrate

ACS reagent, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOH · H2O
CAS Number:
Molecular Weight:
41.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

Assay

≥98.0%

form

powder, crystals or granules

impurities

≤0.01% insolubles
≤2.0% Li2CO3

pH

12 (0.4 g/L)

solubility

water: soluble 216 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.05%

cation traces

Fe: ≤0.002%
heavy metals: ≤20 ppm (by ICP-OES)

storage temp.

room temp

SMILES string

[Li+].O.[OH-]

InChI

1S/Li.2H2O/h;2*1H2/q+1;;/p-1

InChI key

GLXDVVHUTZTUQK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hydroxide monohydrate is a lithium salt. Crystal structure study reveals it has monoclinic structure with space group C2/m. Its vibrational spectra has been studied using IR, Raman and inelastic neutron scattering (INS) spectroscopy. It is reported to transform the surface of titania monoliths to form flower like structures.

Application

Lithium hydroxide monohydrate may be used to produce naphtoxy ion during the synthesis of allyl 1-naphthyl ethers.
It may be used in the synthesis of the following:
  • Lithium titanate (Li4Ti5O12) heterogeneous nanostructures.
  • Tin oxide-carbon (SnO2/C) nanocomposites.
  • Zinc oxide (ZnO) nanowire arrays.
  • Lithium diborate gel films.
  • Porous layered lithium cobalt oxide (LiCoO2) films.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of heterogeneous Li4Ti5O12 nanostructured anodes with long-term cycle stability.
Lee DK, et al.
Nanoscale Research Letters, 5(10), 1585-1589 (2010)
Flower-like surface modification of titania materials by lithium hydroxide solution.
Hasegawa G, et al.
Journal of Colloid and Interface Science, 374(1), 291-296 (2012)
Optimization of the ultrasound-assisted synthesis of allyl 1-naphthyl ether using response surface methodology.
dos Santos Fernandes JP, et al.
Ultrasonics Sonochemistry, 18(2), 489-493 (2011)
Lithium borate gel films prepared from aqueous solution.
Hemry JD, et al.
J. Mater. Sci., 33(15), 3853-3858 (1998)
Al-doped ZnO thin film: a new transparent conducting layer for ZnO nanowire-based dye-sensitized solar cells.
Lee SH, et al.
The Journal of Physical Chemistry C, 114(15), 7185-7189 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service