Skip to Content
Merck
All Photos(3)

Key Documents

399124

Sigma-Aldrich

Potassium permanganate

ACS reagent, ≥99.0%, low in mercury

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KMnO4
CAS Number:
Molecular Weight:
158.03
EC Number:
MDL number:
UNSPSC Code:
12352312
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0%
grade:
ACS reagent
form:
solid
solubility:
water: soluble 63.8 g/L at 20 °C(lit.)
acetone: soluble(lit.)
methanol: soluble(lit.)

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

solid

reaction suitability

reagent type: oxidant

impurities

≤0.005% N compounds
≤0.2% insolubles

pH

7.2-9.7 (20 °C, 20 g/L)

solubility

water: soluble 63.8 g/L at 20 °C(lit.)
acetone: soluble(lit.)
methanol: soluble(lit.)

anion traces

chloride, chlorate (as Cl-): ≤0.005%
sulfate (SO42-): ≤0.02%

cation traces

Hg: ≤0.05 ppm

SMILES string

[K+].[O-][Mn](=O)(=O)=O

InChI

1S/K.Mn.4O/q+1;;;;;-1

InChI key

VZJVWSHVAAUDKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium permanganate (PP, KMnO4) is a dark violet colored crystalline solid. It is a strong oxidizing agent used in the oxidation of a wide range of organic compounds. PP is commonly used as a disinfectant to treat external infections in aquaculture. Corncobs pretreated with alkaline potassium permanganate solution showed high carbohydrate recovery due to the increased enzymatic hydrolysis of lignocellulose. The acidic potassium permanganate chemiluminescence detection has been employed for the detection of cannabinoids.

Application

Potassium permanganate was used in the preparation of layered MnO2 which may be employed as cathodic material in lithium secondary batteries.
It may be used as an oxidant in:
  • The reduction of polycyclic aromatic hydrocarbons found in coal tar pitches (CTPs).
  • Oxidation of decabromodiphenyl ether in sulfuric acid medium.
  • Oxidative cleavage of oximes to the corresponding carbonyl compounds potassium permanganate supported on kieselguhr.
  • Oxidation of progestagens.
Suitable for heterogeneous oxidations.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An environmentally benign method for oxidation of oximes with potassium permanganate supported on kieselguhr under solvent-free conditions.
Zhu LY, et al.
Research on Chemical Intermediates, 39(9), 4315-4320 (2013)
Oxidative Degradation of Decabromodiphenyl Ether (BDE 209) by Potassium Permanganate: Reaction Pathways, Kinetics, and Mechanisms Assisted by Density Functional Theory Calculations.
Shi J, et al.
Environmental Science & Technology, 49(7), 4209-4217 (2015)
Degradation of progestagens by oxidation with potassium permanganate in wastewater effluents.
Fayad PB, et al.
Chemistry Central Journal, 7, 84-84 (2013)
Screening of cannabinoids in industrial-grade hemp using two-dimensional liquid chromatography coupled with acidic potassium permanganate chemiluminescence detection.
Pandohee J, et al.
Journal of Separation Science, 38(12), 2024-2032 (2015)
Potassium Permanganate

Lee DG, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service