Skip to Content
Merck
All Photos(4)

Key Documents

34873

Sigma-Aldrich

Heptane

greener alternative
Heptane

suitable for HPLC, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.06
Assay:
≥99%
technique(s):
HPLC: suitable
bp:
98 °C (lit.)
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

Quality Level

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

Assay

≥99%

form

liquid

autoignition temp.

433 °F

purified by

filtration through 0.2μm

expl. lim.

7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable

impurities

≤0.001% free acid (as CH3COOH)
≤0.01% water (Karl Fischer)
≤1 ppb fluorescence (quinine) at 254 nm
≤1 ppb fluorescence (quinine) at 365 nm

evapn. residue

≤0.0003%

color

APHA: ≤10

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

UV absorption

λ: 200 nm Amax: ≤0.70
λ: 210 nm Amax: ≤0.40
λ: 220 nm Amax: ≤0.10
λ: 230 nm Amax: ≤0.05
λ: 245 nm Amax: ≤0.01
λ: 260 nm Amax: ≤0.01
λ: 400 nm Amax: ≤0.01

greener alternative category

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane for chromatographic purification and thus has been enhanced for "Safer Solvents and Auxiliaries". Click here for more information.
Tools and techniques for solvent selection: green solvent selection guides.

Heptane (n-Heptane) is a linear hydrocarbon. Its cetane number has been reported to be 56.[1]

Application

Greener alternatives to Hexane in HPLC.
Heptane (n-heptane) may be used to compose the solvent mixtures for the purification of N-acylpyrrolidine derivatives of fatty acid methyl esters (FAMEs) by TLC (Thin Layer Chromatography).[2]

Features and Benefits

Greener alternative for hexane

Other Notes

Important notice
The article number 34873-4X2.5L will be discontinued. Please order the single bottle 34873-2.5L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bacterial phospholipid molecular species analysis by ion-pair reversed-phase HPLC/ESI/MS.
Mazzella N, et al.
Journal of Lipid Research, 45(7), 1355-1363 (2004)
A comprehensive modeling study of n-heptane oxidation.
Curran HJ, et al.
Combustion and Flame, 114(1), 149-177 (1998)
Arun K Kota et al.
Advanced materials (Deerfield Beach, Fla.), 24(43), 5838-5843 (2012-08-30)
Hierarchically structured, superoleophobic surfaces are demonstrated that display one of the lowest contact angle hysteresis values ever reported - even with extremely low-surface-tension liquids such as n-heptane. Consequently, these surfaces allow, for the first time, even ≈2 μL n-heptane droplets
Bing Chen et al.
Journal of hazardous materials, 188(1-3), 369-374 (2011-02-26)
The influence of the initial fuel temperature on the burning behavior of n-heptane pool fire was experimentally studied at the State Key Laboratory of Fire Science (SKLFS) large test hall. Circular pool fires with diameters of 100mm, 141 mm, and
Thushara Karunasekara et al.
Journal of chromatography. A, 1218(6), 809-816 (2011-01-07)
Partition coefficients for varied compounds were determined for the organic solvent-propylene carbonate biphasic partition system where the organic solvent is n-heptane, isopentyl ether or 1-octanol. These partition coefficient databases are analyzed using the solvation parameter model facilitating a quantitative comparison

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service