Skip to Content
Merck
All Photos(1)

Documents

338443

Sigma-Aldrich

Sodium silicate solution

reagent grade

Synonym(s):

Sodium trisilicate solution, Water glass

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2O(SiO2)x · xH2O
UNSPSC Code:
12352300
NACRES:
NA.55

grade

reagent grade

Quality Level

form

liquid

composition

Na2O, ~10.6%
SiO2, ~26.5%

concentration

12.0-13.0% Si (gravimetric)
13.4-14.4% NaOH (titration by HCl, titration)

technique(s)

ion chromatography: suitable

density

1.39 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Employed in preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption and catalysis.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Liang Lyu et al.
Food chemistry, 289, 369-376 (2019-04-09)
The effects of postharvest treatment with sodium silicate (Si) (100 mM) on mitochondrial ROS production and energy metabolism of the muskmelon fruits (cv. Yujinxiang) on development of defense responses to Trichothecium roseum were studied. Si treatment decreased decay severity of inoculated
A. M. Healey et al.
Inorganic chemistry, 38(3), 455-458 (2001-10-25)
The structure of NaZnSiO(3)OH, synthesized hydrothermally by reaction of Na(2)ZnSiO(4) and NaOH, has been determined from single-crystal X-ray and powder neutron diffraction data (orthorhombic, space group P2(1)2(1)2(1,) a = 7.6872(2) Å, b = 9.3899(2) Å, c = 5.155(1) Å, Z

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service