Skip to Content
Merck
All Photos(1)

Key Documents

320315

Sigma-Aldrich

Hexane, mixture of isomers

≥98.5%, ACS reagent

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.06
grade:
ACS reagent
bp:
68-70 °C (lit.)
Pricing and availability is not currently available.

Product Name

Hexane, mixture of isomers, ACS reagent, ≥98.5%

grade

ACS reagent

vapor density

3 (vs air)

description

Mixture of hexane isomers plus methylcyclopentane

Assay

≥98.5%

form

liquid

autoignition temp.

437 °F

expl. lim.

7.5 %

dilution

(for analytical testing)

impurities

Thiophene, passes test
≤0.0003 meq/g water-soluble titr. acid
≤0.005% S compounds

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

density

0.672 g/mL at 25 °C (lit.)
0.672 g/mL at 25 °C

SMILES string

C(CCC)(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Contains thiophene

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F

Flash Point(C)

-23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of hexane isomers in a metal-organic framework with triangular channels.
Herm ZR, et al.
Science, 340(6135), 960-964 (2013)
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)
Ali Mohraz et al.
Langmuir : the ACS journal of surfaces and colloids, 21(12), 5298-5306 (2005-06-01)
The development of model materials and image processing methods to directly visualize and quantify colloidal rod assembly by means of confocal laser scanning microscopy (CLSM) is reported. Monodisperse fluorescent colloidal rods are prepared by the uniaxial extensional deformation of sterically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service