288772
Silicic acid
99.9%, 20 μm, purified by refining
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352301
PubChem Substance ID:
NACRES:
SB.52
Recommended Products
Quality Level
Assay
99.9%
form
powder
purified by
refining
particle size
20 μm
SMILES string
O[Si](O)=O
InChI
1S/H2O3Si/c1-4(2)3/h1-2H
InChI key
IJKVHSBPTUYDLN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mitra Shabanian et al.
Oral health & preventive dentistry, 10(1), 59-64 (2012-08-22)
To evaluate the changes in the enamel surface microhardness following the application of various suspensions of Crest and Pooneh toothpastes with and without fluoride. Fifty-six enamel blocks of primary incisors were exposed to a pH-cycling regime consisting of demineralisation and
C Saravanan et al.
Current drug delivery, 9(6), 583-587 (2012-03-29)
The bone infections are the most divesting complications confronted by the physicians and patients. The antibiotic loaded bone cements, such as synthetic ceramics and natural ceramics were accepted for bone infections. The currently available bone cements were not accepted due
Tsuyoshi Yamamoto et al.
Journal of plant research, 125(6), 771-779 (2012-04-25)
Rice (Oryza sativa L.) is a typical Si-accumulating plant and is able to accumulate Si up to >10 % of shoot dry weight. The cell wall has been reported to become thicker under Si-deficient condition. To clarify the relationship between Si
T I Nikoforova
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury, (3)(3), 16-21 (2013-07-04)
This article is focused on the benefits of drug-free therapeutic technologies, such as the use of artificial general siliceous baths for the treatment of patients with arterial hypertension. This approach results in the anti-hypertensive effect simultaneous with the correction of
Paul Curnow et al.
Biochemistry, 51(18), 3776-3785 (2012-04-26)
The synthesis and manipulation of silicon materials on the nanoscale are core themes in nanotechnology research. Inspiration is increasingly being taken from the natural world because the biological mineralization of silicon results in precisely controlled, complex silica structures with dimensions
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service