Skip to Content
Merck
All Photos(1)

Key Documents

25059

Sigma-Aldrich

Potassium acetate

puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOK
CAS Number:
Molecular Weight:
98.14
Beilstein:
3595449
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

<0.0000001 hPa ( 25 °C)

Quality Level

grade

puriss.

Assay

99-101%

form

crystalline

quality

meets analytical specification of Ph. Eur., BP, E261

impurities

residual solvents, complies
≤0.0004% heavy metals (as Pb)
≤0.05% KMnO4 red. matter (as HCOOH)

loss

≤1% loss on drying, 150 °C, 2 h

pH

7.5-9 (20 °C, 5%)

density

1.57 g/cm3 at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤2 mg/kg
Ca: ≤50 mg/kg
Cu: ≤4 mg/kg
Fe: ≤10 mg/kg
Mg: ≤30 mg/kg
Na: ≤5000 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg

SMILES string

[K+].CC([O-])=O

InChI

1S/C2H4O2.K/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

SCVFZCLFOSHCOH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Potassium acetate is a hygroscopic potassium salt. It can be prepared by the neutralization of acetic acid with potassium hydroxide or potassium carbonate. It can be used as nucleophilic catalyst for polymerization and addition reactions, source of acetate anion in substitution and addition reaction. It can be used as a base and promoter in various organic reactions.

Application

Potassium acetate is used as a catalyst:
  • In the preparation of 2-hexadecylfuran from furfuraldehyde and palmitic anhydride.
  • Potassium acetate can be used as a base:,In Lossen rearrangement of hydroxamic acids to carbamic acids.
  • In E2 elimination reactions.
  • In the formation of lactones from alkoxyamide substrates.

Potassium acetate is also used as a promoter:
  • In the borylation of aryl chlorides with bis(pinacolato)diboron in presence of palladium catalyst.
  • In the β-boration of α, β-unsaturated ketones.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 886-886 (1994)
Saša Haberl et al.
The Journal of membrane biology, 246(11), 861-867 (2013-07-09)
The use of plasmid DNA (pDNA) as a pharmaceutical tool has increased since it represents a safer vector for gene transfer compared to viral vectors. Different pDNA extraction methods have been described; among them is alkaline lysis, currently the most
Jay D Gralla et al.
The EMBO journal, 25(7), 1515-1521 (2006-03-17)
Potassium glutamate accumulates upon hyper-osmotic shock and serves as a temporary osmoprotectant. This salt leads to transcriptional activation of sets of genes that allow the cell to achieve long-term adaptation to high osmolarity. The current experiments show that potassium glutamate
Yuval Erez et al.
The journal of physical chemistry. A, 116(9), 2039-2048 (2012-02-10)
Steady-state and time-resolved emission techniques were employed to study the effect of acetate, a mild base, on the luminescence of curcumin in methanol and ethanol. We found that the steady-state emission intensity as well as the average fluorescence decay time
Fátima Gebauer et al.
Methods in enzymology, 429, 23-33 (2007-10-05)
Classically, Drosophila cell-free translation systems have been used to study the response of the translational machinery to heat shock treatment. We and others have developed optimized Drosophila embryo and ovary extracts, and their use has expanded to the study of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service