Skip to Content
Merck
All Photos(2)

Key Documents

210021

Sigma-Aldrich

Hydriodic acid

57 wt. % in H2O, distilled, stabilized, 99.95%

Synonym(s):

Hydriotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HI
CAS Number:
Molecular Weight:
127.91
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

Assay

99.95%

form

liquid

contains

<1.5% hypophosphorous acid as stabilizer

concentration

57 wt. % in H2O

bp

127 °C (lit.)

density

1.701 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

I

InChI

1S/HI/h1H

InChI key

XMBWDFGMSWQBCA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydriodic acid (HI) along with red phosphorus (HI/red P) plays the role of a reducing agent for the preparation of methamphetamine. HI/Phosphorous causes the reduction of various polycyclic quinones. HI also participates as a reducing agent for the conversion of various polyarene quinones, hydroquinones and phenols to the corresponding aromatic hydrocarbons.
Hydriodic acid is hydrogen iodide gas dissolved in water. It is a strong acid generally used as a reducing agent.

Application

Hydriodic acid may be used as a reagent for the conversion of fatty alcohols and glyceryl ethers to alkyl iodides.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the composition of glyceryl ethers and their preparation from diacyl glyceryl ethers in liver oils.
Guyer KE, et al.
Journal of Lipid Research, 4(4), 385-391 (1963)
Chao Chen et al.
Nature communications, 10(1), 1927-1927 (2019-04-28)
Circularly polarized light (CPL) detection is required in various fields such as drug screening, security surveillance and quantum optics. Conventionally, CPL photodetector needs the installation of optical elements, imposing difficulties for integrated and flexible devices. The established CPL detectors without
Efficient reduction of polycyclic quinones, hydroquinones, and phenols with hydriodic acid.
Konieczny M and Harvey RG.
The Journal of Organic Chemistry, 44(26), 4813-4816 (1979)
Methamphetamine synthesis via hydriodic acid/red phosphorus reduction of ephedrine.
Skinner HF.
Forensic Science International, 48(2), 123-134 (1990)
Daniel J Weisenberger et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 24(3), 512-519 (2015-01-15)
The CpG island methylator phenotype (CIMP) represents a subset of colorectal cancers characterized by widespread aberrant DNA hypermethylation at select CpG islands. The risk factors and environmental exposures contributing to etiologic heterogeneity between CIMP and non-CIMP tumors are not known.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service